Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00464703_m1
          See other PSMC3IP GE Assays ›
          Gene Symbol:
          Psmc3ip
          Gene Name:
          proteasome (prosome, macropain) 26S subunit, ATPase 3, interacting protein
          Gene Aliases:
          C79099, GT198, HOP2, Tbpip
          Chromosome Location:
          Chr.11: 101092141 - 101095435 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00464703_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psmc3ip
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008949.3 NP_032975.1 4 - 5 399 57
          GenBank mRNA AB000121.1 - 4 - 5 364 57
          AK133462.1 - 4 - 5 367 57
          BC030169.1 - 4 - 5 376 5027493 57
          FJ937966.1 - 4 - 5 370 57
          FJ937967.1 - 3 - 4 434 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003792.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the FJ937967.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psmc3ip

          Panther Classification:

          Molecular Function -

          DNA metabolism protein nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA recombination
          positive regulation of transcription from RNA polymerase II promoter
          meiotic cell cycle
          regulation of nucleic acid-templated transcription
          DNA binding
          estrogen receptor binding
          ligand-dependent nuclear receptor transcription coactivator activity
          glucocorticoid receptor binding
          protein homodimerization activity
          thyroid hormone receptor binding
          androgen receptor binding
          DBD domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28071925 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28071924 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28071926 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P238044 NM_008949.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P238044 AB000121.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P238044 AK133462.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P238044 BC030169.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P238044 FJ937966.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P238044 NM_008949.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P238044 AB000121.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P238044 AK133462.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P238044 BC030169.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P238044 FJ937966.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 971928 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487725 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142247 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P238044 NM_008949.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P238044 AB000121.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P238044 AK133462.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P238044 BC030169.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P238044 FJ937966.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P238044 NM_008949.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P238044 AB000121.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P238044 AK133462.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P238044 BC030169.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P238044 FJ937966.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28071920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28071915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28071909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28071924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28071929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28071912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28071926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28071908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28071927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28071921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28071928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28071930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28071918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28071917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28071919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28071911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28071923 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142247 - -
          Whole Mouse Genome G4122A A_51_P256706 NM_008949.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256706 AB000121.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256706 AK133462.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256706 BC030169.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256706 FJ937966.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P238044 NM_008949.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P238044 AB000121.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P238044 AK133462.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P238044 BC030169.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P238044 FJ937966.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P238044 NM_008949.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P238044 AB000121.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P238044 AK133462.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P238044 BC030169.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P238044 FJ937966.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110192 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142247 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09640988 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09641001 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09640992 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09641000 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28071926 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28071925 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28071924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28071877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28071905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28071915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09546249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28071903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28071926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28071912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28071917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28071930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28071920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09707514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28071928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28071908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09707520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28071923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09640990 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10391344 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391345 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391347 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391341 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391346 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391342 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391343 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391340 - -
          Affymetrix HT MG-430 PM Array Plate 1425271_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1442058_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10391339 - -
          Mouse Genome 430 2.0 Array 1442058_PM_s_at - -
          Mouse Genome 430 2.0 Array 1425271_PM_at - -
          Mouse Genome 430 2.0 Array 1425271_at - -
          Mouse Genome 430 2.0 Array 1442058_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19183_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19183_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100035023.mm.1 NM_008949.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035023.mm.1 NM_008949.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035023.mm.1 TC1100003792.mm.1 MTA TC
          Mouse Expression Set 430 1425271_PM_at - -
          Mouse Expression Set 430 1425271_at - -
          Mouse Expression Set 430 1442058_s_at - -
          Mouse Expression Set 430 1442058_PM_s_at - -
          Murine Genome U74v2 Set 102970_at - -
          Murine Genome U74v2 Set 102970_PM_at - -
          Mouse Genome 430A 2.0 Array 1425271_at - -
          Mouse Genome 430A 2.0 Array 1425271_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425271_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425271_at - -
          Affymetrix HT Mouse Genome 430B Array 1442058_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1442058_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 920120 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1770039 - -
          Illumina mouse-6 v1.0 expression beadchip 1770039 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001770039 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1237383 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237383 - -
          mouse-6 v1.1 (Illumina) scl39544.7.1_101-S - -
          Illumina Sentrix mouse-6.v1 bead array 1770039 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl39544.7.1_101-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1770039 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2999654 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2999654 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl39544.7.1_101-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline