Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00468054_m1
          See other CD300A GE Assays ›
          Gene Symbol:
          Cd300a
          Gene Name:
          CD300A antigen
          Gene Aliases:
          B230315M08Rik, Clm8, LMIR1, MAIR-I, MAIR-Ia, MMAC8, Pigr4, mcpir1
          Chromosome Location:
          Chr.11: 114889149 - 114904651 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00468054_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd300a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_170758.3 NP_739564.1 3 - 4 872 91
          GenBank mRNA AB065156.1 - 3 - 4 591 91
          AB091765.1 - 3 - 4 598 91
          AB091766.1 - 3 - 4 594 91
          AB095675.1 - 3 - 4 695 91
          AK045869.1 - 3 - 4 872 91
          AK053160.1 - 3 - 4 548 91
          AY457054.1 - 3 - 4 548 91
          BC139296.1 - 3 - 4 808 8862318 91
          BC139297.1 - 3 - 4 808 8862319 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001882.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd300a

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          serotonin secretion by mast cell
          receptor-mediated endocytosis
          signal transduction
          negative regulation of serotonin secretion
          negative regulation of B cell proliferation
          positive regulation of phosphoprotein phosphatase activity
          negative regulation of mast cell activation involved in immune response
          negative regulation of mast cell degranulation
          negative regulation of MAP kinase activity
          negative regulation of fibroblast proliferation
          regulation of T cell receptor signaling pathway
          negative regulation of B cell receptor signaling pathway
          negative regulation of NK T cell activation
          negative regulation of phagocytosis, engulfment
          negative regulation of neutrophil activation
          negative regulation of eosinophil activation
          negative regulation of activation of JAK2 kinase activity
          negative regulation of eosinophil migration
          phosphatidylserine binding
          protein binding
          phosphatidylethanolamine binding
          phosphatase binding
          signaling receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2008297 AK045869.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2008297 NM_170758.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 876914 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 624490 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 246402 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 543103 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 758406 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 392937 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 335121 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 524811 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 131470 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141292 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168009 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107064 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2008297 AK045869.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2008297 NM_170758.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150346 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150319 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107064 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168009 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141292 - -
          Whole Mouse Genome G4122A A_52_P492142 BC139297.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P492142 AY457054.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P492142 BC139296.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P492142 AB091765.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P492142 AB091766.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P147003 AK045869.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P147003 NM_170758.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126927 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125501 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107064 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141292 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168009 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09587527 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09587519 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09587532 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09587520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09554529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09587529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09587532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150376 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09704598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09122794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09420896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09543738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09195215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09477239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150381 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150385 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09122793 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10382447 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382446 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382445 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382439 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382444 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382441 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382440 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382442 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382443 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382448 - -
          Affymetrix HT MG-430 PM Array Plate 1435903_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1445292_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10382438 - -
          Mouse Genome 430 2.0 Array 1445292_PM_at - -
          Mouse Genome 430 2.0 Array 1445292_at - -
          Mouse Genome 430 2.0 Array 1435903_PM_at - -
          Mouse Genome 430 2.0 Array 1435903_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 217303_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 217303_at - -
          Affymetrix Mouse Clariom S Array PSR1100017644.mm.1 NM_170758.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017644.mm.1 NM_170758.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017644.mm.1 TC1100001882.mm.1 MTA TC
          Mouse Expression Set 430 1435903_at - -
          Mouse Expression Set 430 1435903_PM_at - -
          Mouse Expression Set 430 1445292_PM_at - -
          Mouse Expression Set 430 1445292_at - -
          Murine Genome U74v2 Set 104023_PM_at - -
          Murine Genome U74v2 Set 104023_at - -
          Mouse Genome 430A 2.0 Array 1435903_at - -
          Mouse Genome 430A 2.0 Array 1435903_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1435903_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1435903_at - -
          Affymetrix HT Mouse Genome 430B Array 1445292_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1445292_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 567873 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6760128 - -
          Illumina mouse-6 v1.0 expression beadchip 103990142 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006760128 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1218797 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218797 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1247685 - -
          mouse-6 v1.1 (Illumina) scl40740.8_132-S - -
          Illumina Sentrix mouse-6.v1 bead array 103990142 - -
          Illumina Sentrix mouse-6.v1 bead array 6760128 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40740.8_132-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6760128 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103990142 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2887983 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2887986 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2887986 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2887983 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1247685 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline