Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00470225_g1
          See other ICAM4 GE Assays ›
          Gene Symbol:
          Icam4
          Gene Name:
          intercellular adhesion molecule 4, Landsteiner-Wiener blood group
          Gene Aliases:
          1810015M19Rik, Cd242, ICAM-4
          Chromosome Location:
          Chr.9: 21029373 - 21030531 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00470225_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Icam4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023892.2 NP_076381.1 2 - 3 759 64
          GenBank mRNA AF296282.1 - 2 - 3 678 64
          AK019013.1 - 2 - 3 461 64
          BC116909.1 - 2 - 3 759 40126228 64
          BC116913.1 - 2 - 3 759 40126232 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000185.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Icam4

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          single organismal cell-cell adhesion
          integrin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235726 BC116909.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235726 BC116913.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235726 NM_023892.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235726 AF296282.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235726 AF296283.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235726 BC116909.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235726 BC116913.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235726 NM_023892.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235726 AF296282.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235726 AF296283.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235726 BC116909.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235726 BC116913.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235726 NM_023892.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235726 AF296282.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235726 AF296283.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235726 BC116909.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235726 BC116913.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235726 NM_023892.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235726 AF296282.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235726 AF296283.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P235726 BC116909.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P235726 BC116913.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P235726 NM_023892.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P235726 AF296282.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P235726 AF296283.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P235726 BC116909.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P235726 BC116913.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P235726 NM_023892.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P235726 AF296282.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P235726 AF296283.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26345126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26345132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26345130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26345133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26345135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26345129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26345127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26345134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26345124 - -
          Whole Mouse Genome G4122A A_51_P235726 BC116909.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235726 BC116913.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235726 NM_023892.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235726 AF296282.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235726 AF296283.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235726 BC116909.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235726 BC116913.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235726 NM_023892.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235726 AF296282.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235726 AF296283.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116449 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07660986 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07669103 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07660992 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07660950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26345146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26345127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07660992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26345135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26345134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26345133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26345130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26345129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07669103 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10583531 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583533 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583532 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583530 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583534 - -
          Affymetrix HT MG-430 PM Array Plate 1422930_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10583529 - -
          Mouse Genome 430 2.0 Array 1422930_at - -
          Mouse Genome 430 2.0 Array 1422930_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78369_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78369_at - -
          Affymetrix Mouse Clariom S Array PSR0900001222.mm.1 NM_023892.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001222.mm.1 NM_023892.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001222.mm.1 TC0900000185.mm.1 MTA TC
          Mouse Expression Set 430 1422930_PM_at - -
          Mouse Expression Set 430 1422930_at - -
          Mouse Genome 430A 2.0 Array 1422930_at - -
          Mouse Genome 430A 2.0 Array 1422930_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422930_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422930_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 848403 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6770053 - -
          MouseWG-6 v2 Expression BeadChip 101570112 - -
          Illumina mouse-6 v1.0 expression beadchip 101570112 - -
          Illumina mouse-6 v1.0 expression beadchip 6770053 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006770053 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2637808 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1253293 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2637808 - -
          mouse-6 v1.1 (Illumina) scl37235.3.1_1-S - -
          mouse-6 v1.1 (Illumina) scl0003449.1_5-S - -
          Illumina Sentrix mouse-6.v1 bead array 6770053 - -
          Illumina Sentrix mouse-6.v1 bead array 101570112 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37235.3.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101570112 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6770053 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2637808 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2637808 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline