Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00471424_m1
          See other ZCCHC10 GE Assays ›
          Gene Symbol:
          Zcchc10
          Gene Name:
          zinc finger, CCHC domain containing 10
          Gene Aliases:
          2410141K03Rik, AA675321, D11Ertd416e
          Chromosome Location:
          Chr.11: 53324689 - 53333301 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00471424_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zcchc10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026479.4 NP_080755.2 3 - 4 258 90
          GenBank mRNA AK010803.1 - 3 - 4 269 90
          AK020177.1 - 3 - 4 259 90
          AK027985.1 - 3 - 4 266 90
          AK169856.1 - 3 - 4 259 90
          BC025078.1 - 3 - 4 274 4504126 90
          BC087902.1 - 3 - 4 258 30505854 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000600.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zcchc10

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          nucleic acid binding
          zinc ion binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27804179 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27804178 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27804180 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P514675 BC025078.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P514675 BC087902.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P514675 AK027985.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P514675 AK169856.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P514675 NM_026479.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P514675 BC025078.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P514675 BC087902.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P514675 AK027985.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P514675 AK169856.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P514675 NM_026479.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 468873 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 783023 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 261961 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 117389 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 203877 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 863112 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 2338 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159164 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P514675 BC025078.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P514675 BC087902.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P514675 AK027985.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P514675 AK169856.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P514675 NM_026479.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P514675 BC025078.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P514675 BC087902.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P514675 AK027985.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P514675 AK169856.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P514675 NM_026479.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27804164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27804165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27804191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27804184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27804176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27804167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27804169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27804188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27804179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27804171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27804186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27804174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27804183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27804180 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27804172 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159164 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07345043 - -
          Whole Mouse Genome G4122A A_51_P514675 BC025078.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P514675 BC087902.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P514675 AK027985.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P514675 AK169856.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P514675 NM_026479.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P514675 BC025078.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P514675 BC087902.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P514675 AK027985.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P514675 AK169856.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P514675 NM_026479.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118876 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159164 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09126139 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09126138 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27804179 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27804180 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27804178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27804208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09200410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09400208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09091282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09242556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09161294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09639073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09723412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27804159 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10375978 - -
          Affymetrix Mouse Gene 1.0 ST Array 10375977 - -
          Affymetrix Mouse Gene 1.0 ST Array 10375976 - -
          Affymetrix Mouse Gene 1.0 ST Array 10375979 - -
          Affymetrix HT MG-430 PM Array Plate 1418820_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10375975 - -
          Mouse Genome 430 2.0 Array 1418820_s_at - -
          Mouse Genome 430 2.0 Array 1418820_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67966_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67966_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100004697.mm.1 NM_026479.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004697.mm.1 NM_026479.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004697.mm.1 TC1100000600.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100004699.mm.1 NM_026479.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004699.mm.1 NM_026479.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004699.mm.1 TC1100000600.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100004698.mm.1 NM_026479.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004698.mm.1 NM_026479.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004698.mm.1 TC1100000600.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100004700.mm.1 NM_026479.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004700.mm.1 NM_026479.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004700.mm.1 TC1100000600.mm.1 MTA TC
          Mouse Expression Set 430 1418820_PM_s_at - -
          Mouse Expression Set 430 1418820_s_at - -
          Murine Genome U74v2 Set 96189_at - -
          Murine Genome U74v2 Set 96189_PM_at - -
          Mouse Genome 430A 2.0 Array 1418820_s_at - -
          Mouse Genome 430A 2.0 Array 1418820_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1418820_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1418820_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 101850082 - -
          Illumina mouse-6 v1.0 expression beadchip 1410131 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001410131 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2764235 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2462622 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2764235 - -
          mouse-6 v1.1 (Illumina) scl0067966.2_186-S - -
          mouse-6 v1.1 (Illumina) scl41570.5_643-S - -
          Illumina Sentrix mouse-6.v1 bead array 1410131 - -
          Illumina Sentrix mouse-6.v1 bead array 101850082 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0067966.2_186-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101850082 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1410131 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2764235 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2764235 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0