Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00471739_m1
          See other ZFP85OS GE Assays ›
          Gene Symbol:
          Zfp85os
          Gene Name:
          zinc finger protein 85, opposite strand
          Gene Aliases:
          4930441O14Rik
          Chromosome Location:
          Chr.13: 67729262 - 67756838 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00471739_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp85os
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027969.1 2 - 3 647 77
          GenBank mRNA AK015755.1 - 2 - 3 315 77
          AK019608.1 - 2 - 3 364 77
          AK134579.1 - 2 - 3 648 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000828.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK015755.1,AK019608.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P108384 AK019608.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P108384 AK019608.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 932915 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 700203 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 311818 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 746859 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 524847 AK015483.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 524847 AK016243.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 931229 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 764871 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 767444 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 740500 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 166695 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 392009 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 164501 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 491879 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 593310 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 65874 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 61348 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 92482 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 132380 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147602 - -
          Whole Mouse Genome G4122A A_51_P108384 AK019608.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P108384 AK019608.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10596270 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10798533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10798532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10438828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109606 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10405933 - -
          Affymetrix Mouse Gene 1.0 ST Array 10405934 - -
          Affymetrix HT MG-430 PM Array Plate 1419335_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10405931 - -
          Mouse Genome 430 2.0 Array 1419335_PM_at - -
          Mouse Genome 430 2.0 Array 1419335_at - -
          Affymetrix Mouse Clariom S Array PSR1300005391.mm.1 NR_027969.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005391.mm.1 NR_027969.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005391.mm.1 TC1300000828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300005393.mm.1 NR_027969.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005393.mm.1 NR_027969.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005393.mm.1 TC1300000828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300005387.mm.1 NR_027969.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005387.mm.1 NR_027969.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005387.mm.1 TC1300000828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300005394.mm.1 NR_027969.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005394.mm.1 NR_027969.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005394.mm.1 TC1300000828.mm.1 MTA TC
          Murine Genome U74v2 Set 170833_at - -
          Murine Genome U74v2 Set 170833_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 441910 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2320563 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline