Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00474870_m1
          See other 1700102P08RIK GE Assays ›
          Gene Symbol:
          1700102P08Rik
          Gene Name:
          RIKEN cDNA 1700102P08 gene
          Gene Aliases:
          AI593206
          Chromosome Location:
          Chr.9: 108392834 - 108410966 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00474870_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700102P08Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053216.2 NP_444446.2 2 - 3 427 93
          GenBank mRNA AK007115.1 - 2 - 3 427 93
          BC061048.1 - 2 - 3 427 6770968 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001404.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 1700102P08Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P382802 AK007115.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P382802 BC061048.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P382802 NM_053216.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P382802 AK007115.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P382802 BC061048.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P382802 NM_053216.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 876061 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 621887 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 329284 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 488632 BC052527.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 117110 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166888 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P382802 AK007115.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P382802 BC061048.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P382802 NM_053216.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P382802 AK007115.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P382802 BC061048.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P382802 NM_053216.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814653 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166888 - -
          Whole Mouse Genome G4122A A_52_P382802 AK007115.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P382802 BC061048.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P382802 NM_053216.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P382802 AK007115.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P382802 BC061048.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P382802 NM_053216.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139201 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166888 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08164774 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08164769 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08164783 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08164772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07538801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08105422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07675593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07538806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07538803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07538802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07763742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07580645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08164773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07955131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08105425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08105404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07580643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07462267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08105426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08059566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814690 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10588930 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588929 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588928 - -
          Affymetrix HT MG-430 PM Array Plate 1419531_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10588927 - -
          Mouse Genome 430 2.0 Array 1419531_at - -
          Mouse Genome 430 2.0 Array 1419531_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 112418_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 112418_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900011279.mm.1 NM_053216.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011279.mm.1 NM_053216.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011279.mm.1 TC0900001404.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900011280.mm.1 NM_053216.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011280.mm.1 NM_053216.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011280.mm.1 TC0900001404.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900011281.mm.1 NM_053216.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011281.mm.1 NM_053216.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011281.mm.1 TC0900001404.mm.1 MTA TC
          Mouse Expression Set 430 1419531_at - -
          Mouse Expression Set 430 1419531_PM_at - -
          Mouse Genome 430A 2.0 Array 1419531_at - -
          Mouse Genome 430A 2.0 Array 1419531_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419531_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419531_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 851333 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5670180 - -
          Illumina mouse-6 v1.0 expression beadchip 5670180 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005670180 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2646254 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2646254 - -
          mouse-6 v1.1 (Illumina) scl36471.3.1_3-S - -
          Illumina Sentrix mouse-6.v1 bead array 5670180 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36471.3.1_3-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5670180 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2646254 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2646254 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline