Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00476483_m1
          See other NPDC1 GE Assays ›
          Gene Symbol:
          Npdc1
          Gene Name:
          neural proliferation, differentiation and control 1
          Gene Aliases:
          AI314472, NPDC-1
          Chromosome Location:
          Chr.2: 25403050 - 25409494 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00476483_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Npdc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008721.4 NP_032747.2 2 - 3 467 87
          GenBank mRNA AK146466.1 - 2 - 3 356 87
          BC003320.1 - 2 - 3 434 3155595 87
          L03814.1 - 2 - 3 271 87
          X67209.1 - 2 - 3 271 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200005463.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Npdc1

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21071912 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21071913 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21071909 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21071914 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21071911 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974650 BC003320.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974650 BC023887.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974650 L03814.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974650 X67209.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974650 NM_008721.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 409532 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 624995 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 235093 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 55865 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120699 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974650 BC003320.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974650 BC023887.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974650 L03814.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974650 X67209.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974650 NM_008721.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071904 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071906 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21071950 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120699 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00324988 - -
          Whole Mouse Genome G4122A A_51_P154596 BC003320.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154596 BC023887.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154596 L03814.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154596 X67209.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154596 NM_008721.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125502 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120699 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01185508 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01742298 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01742292 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01185504 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01474052 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01742290 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01742288 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01742287 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01325263 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01185511 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01185516 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01742291 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01474053 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21071911 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21071914 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21071913 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21071909 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21071912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01340675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02229129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02229126 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01321055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071967 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01833709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071941 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01742289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071923 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071969 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01716681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071942 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01833694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02117599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071901 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071961 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071958 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01185508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071960 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071971 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071964 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01833700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01922787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01340680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01742294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01325263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02229130 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10470039 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470038 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470031 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470033 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470032 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470040 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470034 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470041 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470030 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470028 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470035 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470036 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470029 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470037 - -
          Affymetrix HT MG-430 PM Array Plate 1415919_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10470027 - -
          Mouse Genome 430 2.0 Array 1415919_PM_at - -
          Mouse Genome 430 2.0 Array 1415919_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18146_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18146_at - -
          Affymetrix Murine Genome 6500A Array Msa10270_at - -
          Affymetrix Mouse Clariom S Array PSR0200046646.mm.1 NM_008721.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200046646.mm.1 TC0200005463.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200002386.mm.1 NM_008721.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002386.mm.1 TC0200005463.mm.1 MTA TC
          Mouse Expression Set 430 1415919_at - -
          Mouse Expression Set 430 1415919_PM_at - -
          Murine Genome U74v2 Set 92626_PM_at - -
          Murine Genome U74v2 Set 92626_at - -
          Mouse Genome 430A 2.0 Array 1415919_at - -
          Mouse Genome 430A 2.0 Array 1415919_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1415919_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1415919_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 690152 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2230047 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002230047 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2589403 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2589403 - -
          mouse-6 v1.1 (Illumina) scl018146.8_16-S - -
          Illumina Sentrix mouse-6.v1 bead array 2230047 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl018146.8_16-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2230047 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2589403 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2589403 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl018146.8_16-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline