| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2122884 |
BC119062.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2122884 |
BC120746.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2122884 |
NM_011195.2 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2122884 |
U16958.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
547475 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
445980 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
759491 |
AF187099.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
759491 |
AK007530.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
759491 |
BC024605.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
759491 |
NM_023493.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
935256 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
911504 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
128014 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
147584 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2122884 |
BC119062.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2122884 |
BC120746.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2122884 |
NM_011195.2 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2122884 |
U16958.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P243575 |
U16958.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208795 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208831 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208817 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208810 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208778 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208800 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208834 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208802 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208771 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208773 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208836 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208799 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208770 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208796 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208805 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208777 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208807 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208813 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208798 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208839 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208833 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208769 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208819 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208838 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208842 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208823 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208820 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208775 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208828 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208837 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208840 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208779 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208826 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208808 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31208768 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P03325376 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P03325368 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P243575 |
U16958.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P128356 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P03325376 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13169581 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13500860 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13225567 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13225560 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13225559 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13225566 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13500859 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13500862 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208805 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208764 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208722 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208781 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208833 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208777 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208785 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208720 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208826 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208732 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208791 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208788 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208846 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208813 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208747 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208759 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208842 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208825 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208782 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208838 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208737 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208717 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208771 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208808 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208819 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208766 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208800 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208745 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208844 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208758 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208749 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208783 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208820 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208798 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208744 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208730 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208741 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208828 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208775 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208762 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208773 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208790 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208748 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208769 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208724 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208756 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208728 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208713 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208770 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208799 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208831 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208752 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208836 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208754 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208807 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208839 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208796 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208834 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208840 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31208848 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31208783 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31208815 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31208777 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31208773 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31208771 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31208805 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13495380 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31208794 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31208781 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13278404 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13411088 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31208782 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31208819 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31208787 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13232002 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31208807 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31208785 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13278411 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31208795 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31208802 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31208798 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31208796 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31208799 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31208810 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13232001 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31208800 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13278401 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31208779 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31208791 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31208770 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31208808 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31208813 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31208767 |
- |
- |