Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00480018_m1
          See other ARL2 GE Assays ›
          Gene Symbol:
          Arl2
          Gene Name:
          ADP-ribosylation factor-like 2
          Gene Aliases:
          2610009M23Rik, AI115441, AW553335
          Chromosome Location:
          Chr.19: 6134389 - 6141137 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00480018_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arl2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019722.3 NP_062696.2 3 - 4 376 72
          GenBank mRNA AF143680.1 - 3 - 4 340 72
          AK011366.1 - 3 - 4 376 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001004.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arl2

          Gene Ontology Categories:

          Function(s) Process(es)

          cell cycle
          small GTPase mediated signal transduction
          acetylcholine transport
          regulation of microtubule polymerization
          positive regulation of microtubule polymerization
          negative regulation of GTPase activity
          centrosome organization
          maintenance of protein location in nucleus
          nucleotide binding
          GTPase activity
          protein binding
          GTP binding
          GDP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932938 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932936 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932937 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932934 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932932 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932931 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P122696 BC080803.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P122696 NM_019722.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P122696 AK011366.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P122696 BC060259.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P122696 BC080803.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P122696 NM_019722.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P122696 AK011366.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P122696 BC060259.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 338704 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 881031 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 278030 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 18137 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108191 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P122696 BC080803.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P122696 NM_019722.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P122696 AK011366.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P122696 BC060259.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P122696 BC080803.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P122696 NM_019722.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P122696 AK011366.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P122696 BC060259.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P373226 AF143680.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P122696 BC080803.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P122696 NM_019722.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P122696 AK011366.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P122696 BC060259.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P122696 BC080803.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P122696 NM_019722.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P122696 AK011366.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P122696 BC060259.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932933 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P108191 - -
          Whole Mouse Genome G4122A A_52_P122696 BC080803.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P122696 NM_019722.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P122696 AK011366.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P122696 BC060259.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P122696 BC080803.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P122696 NM_019722.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P122696 AK011366.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P122696 BC060259.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P373226 AF143680.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126967 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128171 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P108191 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14453689 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14453693 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14453692 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14453686 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14453690 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932937 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932932 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932936 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932934 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932938 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14409578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14405705 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14456357 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14453686 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14453689 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932946 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14474565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14464826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14285426 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14408852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932902 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932960 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14298134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14464823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14298135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14435033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14408857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932923 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14307588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14270284 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10465390 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465393 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465392 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465389 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465394 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465391 - -
          Affymetrix HT MG-430 PM Array Plate 1422549_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1458565_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10465388 - -
          Mouse Genome 430 2.0 Array 1458565_at - -
          Mouse Genome 430 2.0 Array 1422549_PM_at - -
          Mouse Genome 430 2.0 Array 1458565_PM_at - -
          Mouse Genome 430 2.0 Array 1422549_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56327_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56327_at - -
          Affymetrix Mouse Clariom S Array PSR1900009290.mm.1 NM_019722.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009290.mm.1 NM_019722.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009290.mm.1 TC1900001004.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009288.mm.1 NM_019722.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009288.mm.1 NM_019722.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009288.mm.1 TC1900001004.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009289.mm.1 NM_019722.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009289.mm.1 NM_019722.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009289.mm.1 TC1900001004.mm.1 MTA TC
          Mouse Expression Set 430 1422549_at - -
          Mouse Expression Set 430 1422549_PM_at - -
          Mouse Expression Set 430 1458565_at - -
          Mouse Expression Set 430 1458565_PM_at - -
          Murine Genome U74v2 Set 103854_at - -
          Murine Genome U74v2 Set 103854_PM_at - -
          Mouse Genome 430A 2.0 Array 1422549_at - -
          Mouse Genome 430A 2.0 Array 1422549_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422549_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422549_at - -
          Affymetrix HT Mouse Genome 430B Array 1458565_at - -
          Affymetrix HT Mouse Genome 430B Array 1458565_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 847838 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5890377 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005890377 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2620304 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2620304 - -
          mouse-6 v1.1 (Illumina) scl53528.4.1_29-S - -
          Illumina Sentrix mouse-6.v1 bead array 5890377 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl53528.4.1_29-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5890377 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2620304 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2620304 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl53528.4.1_29-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0