Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00480494_m1
          See other AP4M1 GE Assays ›
          Gene Symbol:
          Ap4m1
          Gene Name:
          adaptor-related protein complex AP-4, mu 1
          Gene Aliases:
          4930443L05Rik, Ap4m4
          Chromosome Location:
          Chr.5: 138172021 - 138178687 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00480494_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ap4m1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021392.4 NP_067367.3 4 - 5 517 76
          GenBank mRNA AF242858.1 - 4 - 5 385 76
          AK031638.1 - 4 - 5 520 76
          BC011174.1 - 4 - 5 402 4207033 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001644.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ap4m1

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s)

          transport
          intracellular protein transport
          Golgi to endosome transport
          protein transport
          vesicle-mediated transport

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129164 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129165 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P393067 AK031638.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P393067 BC011174.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P393067 BE627979.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P393067 NM_021392.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P393067 AF242858.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P393067 AK031638.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P393067 BC011174.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P393067 BE627979.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P393067 NM_021392.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P393067 AF242858.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 201015 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 887626 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 630090 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 530846 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 5711 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107386 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P393067 AK031638.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P393067 BC011174.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P393067 BE627979.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P393067 NM_021392.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P393067 AF242858.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P393067 AK031638.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P393067 BC011174.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P393067 BE627979.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P393067 NM_021392.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P393067 AF242858.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129177 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129192 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107386 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01232697 - -
          Whole Mouse Genome G4122A A_51_P393067 AK031638.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P393067 BC011174.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P393067 BE627979.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P393067 NM_021392.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P393067 AF242858.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P393067 AK031638.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P393067 BC011174.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P393067 BE627979.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P393067 NM_021392.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P393067 AF242858.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116160 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107386 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04792853 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04792834 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04792859 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04792845 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04792847 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129165 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129174 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04716252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04685820 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04716254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129166 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129208 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04716257 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04477875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129202 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129200 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129192 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129177 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04716253 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129189 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04817125 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04792860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04865049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129170 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04865051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129172 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10526764 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526755 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526769 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526758 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526762 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526763 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526766 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526756 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526761 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526759 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526771 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526767 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526760 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526768 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526757 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526770 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526765 - -
          Affymetrix HT MG-430 PM Array Plate 1418846_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10526754 - -
          Mouse Genome 430 2.0 Array 1418846_PM_at - -
          Mouse Genome 430 2.0 Array 1418846_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11781_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11781_at - -
          Affymetrix Mouse Clariom S Array PSR0500015072.mm.1 TC0500001644.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015072.mm.1 NM_021392.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015072.mm.1 NM_021392.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015073.mm.1 TC0500001644.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015073.mm.1 NM_021392.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015073.mm.1 NM_021392.4 Refseq
          Mouse Expression Set 430 1418846_PM_at - -
          Mouse Expression Set 430 1418846_at - -
          Mouse Genome 430A 2.0 Array 1418846_at - -
          Mouse Genome 430A 2.0 Array 1418846_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418846_at - -
          Affymetrix HT Mouse Genome 430A Array 1418846_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 363752 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 70609 - -
          Illumina mouse-6 v1.0 expression beadchip 70609 - -
          Illumina mouse-6 v1.0 expression beadchip 3440128 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000070609 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003440128 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2692762 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2599667 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2692762 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2599667 - -
          mouse-6 v1.1 (Illumina) scl27083.14.1_96-S - -
          mouse-6 v1.1 (Illumina) scl0004006.1_26-S - -
          Illumina Sentrix mouse-6.v1 bead array 70609 - -
          Illumina Sentrix mouse-6.v1 bead array 3440128 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0004006.1_26-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl27083.14.1_96-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 70609 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3440128 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2692762 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2856646 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2599667 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2599667 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2692762 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2856646 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl27083.14.1_96-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0