Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00481985_m1
          See other 4933400A11RIK GE Assays ›
          Gene Symbol:
          4933400A11Rik
          Gene Name:
          capping protein (actin filament) muscle Z-line, alpha 1 pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 169776378 - 169779635 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00481985_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4933400A11Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003635.1 2 - 3 204 109
          GenBank mRNA AK016585.1 - 2 - 3 205 109
          BC107241.1 - 2 - 3 126 109

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00003378.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK016585.1 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P127223 AK016585.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P127223 NR_003635.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2099770 BC107240.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2099770 BC107241.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2099770 AK016585.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2099770 NR_003635.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2099770 AK015979.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2099770 BC107240.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2099770 BC107241.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2099770 AK016585.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2099770 NR_003635.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2099770 AK015979.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P127223 AK016585.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P127223 NR_003635.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P662899 BC107240.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P662899 BC107241.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P662899 AK016585.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P662899 NR_003635.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P662899 AK015979.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10607949 - -
          Affymetrix Mouse Gene 1.0 ST Array 10607948 - -
          Affymetrix Mouse Gene 1.0 ST Array 10607947 - -
          Affymetrix HT MG-430 PM Array Plate 1420777_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10607945 - -
          Mouse Genome 430 2.0 Array 1420777_PM_a_at - -
          Mouse Genome 430 2.0 Array 1420777_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66747_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66747_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00021093.mm.1 NR_003635.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00021093.mm.1 TC0X00003378.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00021094.mm.1 NR_003635.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00021094.mm.1 TC0X00003378.mm.1 MTA TC
          Mouse Expression Set 430 1420777_a_at - -
          Mouse Expression Set 430 1420777_PM_a_at - -
          Murine Genome U74v2 Set 167960_r_at - -
          Murine Genome U74v2 Set 167960_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1420777_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1420777_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1420777_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1420777_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 821304 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6900139 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1250324 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1250324 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1250324 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1250324 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline