Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00482278_m1
          See other 2310030G06RIK GE Assays ›
          Gene Symbol:
          2310030G06Rik
          Gene Name:
          RIKEN cDNA 2310030G06 gene
          Gene Aliases:
          2610024D04Rik, AV017521
          Chromosome Location:
          Chr.9: 50739691 - 50746521 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00482278_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2310030G06Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025865.2 NP_080141.1 1 - 2 85 81
          GenBank mRNA AK007611.1 - 1 - 2 101 81
          AK007929.1 - 1 - 2 74 81
          AK009541.1 - 1 - 2 92 81
          BC027409.1 - 1 - 2 85 4035256 81

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK007929.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 2310030G06Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P227165 BC027409.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P227165 AK009541.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P227165 NM_025865.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P227165 BC027409.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P227165 AK009541.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P227165 NM_025865.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 573650 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 924776 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 415411 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 86599 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134104 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P227165 BC027409.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P227165 AK009541.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P227165 NM_025865.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P227165 BC027409.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P227165 AK009541.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P227165 NM_025865.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505410 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505397 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505411 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505383 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505382 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505413 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505388 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505381 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505407 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505406 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505377 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505380 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505390 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505395 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505386 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505375 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505399 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505409 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505400 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134104 - -
          Whole Mouse Genome G4122A A_51_P227165 BC027409.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P227165 AK009541.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P227165 NM_025865.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P227165 BC027409.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P227165 AK009541.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P227165 NM_025865.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131741 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134104 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07955457 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07955454 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08039203 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07955449 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07955455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505352 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505381 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07507802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07571404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07507804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07629797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07571405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505397 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07629798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07791267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08153570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07709815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07507826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07507808 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08153569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07846760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505325 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10593416 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593417 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593415 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593414 - -
          Affymetrix HT MG-430 PM Array Plate 1449357_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10593413 - -
          Mouse Genome 430 2.0 Array 1449357_at - -
          Mouse Genome 430 2.0 Array 1449357_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66952_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66952_at - -
          Affymetrix Mouse Clariom S Array PSR0900018983.mm.1 NM_025865.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900018983.mm.1 NM_025865.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900018983.mm.1 TC0900002276.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900018985.mm.1 NM_025865.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900018985.mm.1 NM_025865.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900018985.mm.1 TC0900002276.mm.1 MTA TC
          Mouse Expression Set 430 1449357_PM_at - -
          Mouse Expression Set 430 1449357_at - -
          Murine Genome U74v2 Set 165630_at - -
          Murine Genome U74v2 Set 165630_PM_at - -
          Murine Genome U74v2 Set 130292_at - -
          Murine Genome U74v2 Set 130292_PM_at - -
          Mouse Genome 430A 2.0 Array 1449357_PM_at - -
          Mouse Genome 430A 2.0 Array 1449357_at - -
          Affymetrix HT Mouse Genome 430A Array 1449357_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449357_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 542766 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6940086 - -
          Illumina mouse-6 v1.0 expression beadchip 6940086 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006940086 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2707595 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2707595 - -
          mouse-6 v1.1 (Illumina) scl35874.4_272-S - -
          Illumina Sentrix mouse-6.v1 bead array 6940086 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35874.4_272-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6940086 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2707595 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2707595 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline