Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00482770_m1
          See other QPCTL GE Assays ›
          Gene Symbol:
          Qpctl
          Gene Name:
          glutaminyl-peptide cyclotransferase-like
          Gene Aliases:
          1810019P04Rik, BB101812, gQC
          Chromosome Location:
          Chr.7: 19140217 - 19149196 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00482770_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Qpctl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026111.3 NP_080387.2 2 - 3 366 59
          GenBank mRNA AK007555.1 - 2 - 3 414 59
          AK007701.1 - 2 - 3 394 59
          AK035587.1 - 2 - 3 366 59
          AK045687.1 - 2 - 3 370 59
          BC058181.1 - 2 - 3 389 5325516 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002471.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Qpctl

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          peptidyl-pyroglutamic acid biosynthetic process, using glutaminyl-peptide cyclotransferase
          zinc ion binding
          glutaminyl-peptide cyclotransferase activity
          transferase activity
          transferase activity, transferring acyl groups
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24991721 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24991723 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24991725 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24991722 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24991726 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24991724 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24991720 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24991719 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P515623 BC058181.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P515623 AK035587.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P515623 AK045687.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P515623 AK136746.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P515623 NM_026111.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P515623 BC058181.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P515623 AK035587.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P515623 AK045687.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P515623 AK136746.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P515623 NM_026111.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 202254 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 529114 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 235119 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 820511 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 7029 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 451937 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P178359 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P515623 BC058181.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P515623 AK035587.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P515623 AK045687.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P515623 AK136746.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P515623 NM_026111.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P515623 BC058181.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P515623 AK035587.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P515623 AK045687.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P515623 AK136746.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P515623 NM_026111.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24991690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24991704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24991698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24991702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24991725 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24991726 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24991694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24991718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24991711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24991719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24991707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24991693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24991721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24991695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24991692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24991724 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24991708 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24991706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24991697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24991713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24991700 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24991716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24991709 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06134300 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06134275 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P178359 - -
          Whole Mouse Genome G4122A A_51_P515623 BC058181.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P515623 AK035587.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P515623 AK045687.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P515623 AK136746.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P515623 NM_026111.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P515623 BC058181.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P515623 AK035587.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P515623 AK045687.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P515623 AK136746.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P515623 NM_026111.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134986 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P178359 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05899959 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05899940 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05899951 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05899944 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05899945 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24991724 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24991719 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24991722 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24991723 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24991721 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24991725 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24991720 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24991726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24991709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24991687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06449866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24991681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24991684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24991721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06449868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24991711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06409957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05899944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24991700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24991718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24991686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06449873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06328654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06328651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24991689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06328655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24991690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24991713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24991716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06550781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24991714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06449871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05909397 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24991685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06451725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24991694 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10560436 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560440 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560442 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560441 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560437 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560435 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560438 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560439 - -
          Affymetrix HT MG-430 PM Array Plate 1449518_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1449517_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10560434 - -
          Mouse Genome 430 2.0 Array 1441323_PM_at - -
          Mouse Genome 430 2.0 Array 1449518_PM_at - -
          Mouse Genome 430 2.0 Array 1449517_PM_at - -
          Mouse Genome 430 2.0 Array 1449517_at - -
          Mouse Genome 430 2.0 Array 1441323_at - -
          Mouse Genome 430 2.0 Array 1449518_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67369_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67369_at - -
          Affymetrix Mouse Clariom S Array PSR0700021736.mm.1 NM_026111.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021736.mm.1 NM_026111.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021736.mm.1 TC0700002471.mm.1 MTA TC
          Mouse Expression Set 430 1449517_at - -
          Mouse Expression Set 430 1449518_PM_at - -
          Mouse Expression Set 430 1449517_PM_at - -
          Mouse Expression Set 430 1449518_at - -
          Mouse Expression Set 430 1441323_at - -
          Mouse Expression Set 430 1441323_PM_at - -
          Murine Genome U74v2 Set 135305_PM_at - -
          Murine Genome U74v2 Set 135305_at - -
          Mouse Genome 430A 2.0 Array 1449517_PM_at - -
          Mouse Genome 430A 2.0 Array 1449518_at - -
          Mouse Genome 430A 2.0 Array 1449517_at - -
          Mouse Genome 430A 2.0 Array 1449518_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449518_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449517_at - -
          Affymetrix HT Mouse Genome 430A Array 1449517_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449518_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 418566 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2810097 - -
          Illumina mouse-6 v1.0 expression beadchip 2810097 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002810097 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1222697 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222697 - -
          mouse-6 v1.1 (Illumina) scl31696.7.1_3-S - -
          Illumina Sentrix mouse-6.v1 bead array 2810097 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31696.7.1_3-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2810097 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1222697 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1222697 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline