Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00484032_g1
          See other CTLA2A GE Assays ›
          Gene Symbol:
          Ctla2a
          Gene Name:
          cytotoxic T lymphocyte-associated protein 2 alpha
          Gene Aliases:
          Ctla-2a
          Chromosome Location:
          Chr.13: 60934155 - 60936625 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00484032_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ctla2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145799.1 NP_001139271.1 3 - 4 345 94
          NM_007796.2 NP_031822.2 3 - 4 383 94
          GenBank mRNA AK005471.1 - 3 - 4 320 94
          AK018801.1 - 3 - 4 338 94
          AK132290.1 - 3 - 4 314 94
          AK167433.1 - 3 - 4 316 94
          AK167664.1 - 3 - 4 317 94
          AY034577.1 - 3 - 4 292 94
          BC028437.1 - 3 - 4 309 1244311 94
          X15591.1 - 3 - 4 292 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002143.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ctla2a

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of protein processing
          regulation of regulatory T cell differentiation
          negative regulation of inflammatory response
          proteolysis involved in cellular protein catabolic process
          molecular_function
          cysteine-type endopeptidase activity
          cysteine-type peptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955656 AY034577.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955656 AK132290.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955656 X15591.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955656 NM_001145799.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955656 NM_007796.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P177344 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955656 AY034577.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955656 AK132290.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955656 X15591.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955656 NM_001145799.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955656 NM_007796.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P180747 AY034578.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P180747 NM_007797.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P180747 X15592.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P180747 AK167433.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P180747 AK167664.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088078 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07896489 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07896467 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07896496 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07896474 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07896486 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07896478 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P177344 - -
          Whole Mouse Genome G4122A A_51_P180747 AY034578.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P180747 NM_007797.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P180747 X15592.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P180747 AK167433.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P180747 AK167664.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139757 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P177344 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07896486 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07896467 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10780859 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10630399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10630399 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10616888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11005238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088076 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10937492 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10874000 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10409878 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409877 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409880 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409879 - -
          Affymetrix HT MG-430 PM Array Plate 1448471_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10409876 - -
          Mouse Genome 430 2.0 Array 1416811_PM_s_at - -
          Mouse Genome 430 2.0 Array 1448471_a_at - -
          Mouse Genome 430 2.0 Array 1448471_PM_a_at - -
          Mouse Genome 430 2.0 Array 1416811_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13024_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13024_at - -
          Affymetrix Mouse Clariom S Array PSR1300013753.mm.1 NM_001145799.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013753.mm.1 NM_001145799.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013753.mm.1 NM_007796.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013753.mm.1 NM_007796.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013753.mm.1 TC1300002143.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013751.mm.1 NM_001145799.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013751.mm.1 NM_001145799.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013751.mm.1 NM_007796.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013751.mm.1 NM_007796.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013751.mm.1 TC1300002143.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013752.mm.1 NM_001145799.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013752.mm.1 NM_001145799.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013752.mm.1 NM_007796.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013752.mm.1 NM_007796.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013752.mm.1 TC1300002143.mm.1 MTA TC
          Mouse Expression Set 430 1448471_PM_a_at - -
          Mouse Expression Set 430 1448471_a_at - -
          Mouse Expression Set 430 1416811_PM_s_at - -
          Mouse Expression Set 430 1416811_s_at - -
          Murine Genome U74v2 Set 162421_r_at - -
          Murine Genome U74v2 Set 96912_PM_s_at - -
          Murine Genome U74v2 Set 96912_s_at - -
          Murine Genome U74v2 Set 162421_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1416811_s_at - -
          Mouse Genome 430A 2.0 Array 1448471_a_at - -
          Mouse Genome 430A 2.0 Array 1448471_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1416811_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1448471_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1448471_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 920918 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6350064 - -
          Illumina mouse-6 v1.0 expression beadchip 6350064 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006350064 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1221739 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1221739 - -
          mouse-6 v1.1 (Illumina) scl013024.1_14-S - -
          Illumina Sentrix mouse-6.v1 bead array 6350064 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl013024.1_14-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6350064 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1221739 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1221739 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline