Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00487934_m1
          See other ROBO3 GE Assays ›
          Gene Symbol:
          Robo3
          Gene Name:
          roundabout homolog 3 (Drosophila)
          Gene Aliases:
          Rbig1, Rig-1, Rig1, Robo3a, Robo3b
          Chromosome Location:
          Chr.9: 37416045 - 37433408 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00487934_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Robo3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164767.1 NP_001158239.1 7 - 8 1334 75
          GenBank mRNA AF060570.1 - 6 - 7 1320 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002050.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          neuron migration
          chemotaxis
          multicellular organism development
          nervous system development
          axon guidance
          axon midline choice point recognition
          cell differentiation
          chemoattraction of axon
          chemorepulsion of axon
          commissural neuron axon guidance
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2144886 AK031746.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2144886 NM_001164767.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2147306 AK079316.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2147306 AF060570.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145587 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121117 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2144886 AK031746.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2144886 NM_001164767.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P481489 AF060570.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P481494 AF060570.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07872981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07628418 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08112908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07960322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07872989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07628428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08028831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07487707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08028829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07598874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08028832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08028835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07695855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07872988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08028846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26430660 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10592326 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592318 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592305 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592312 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592316 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592308 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592324 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592320 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592325 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592321 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592304 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592322 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592311 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592323 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592328 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592314 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592327 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592306 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592310 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592307 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592309 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592317 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592313 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592315 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592319 - -
          Affymetrix HT MG-430 PM Array Plate 1421565_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1436634_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10592303 - -
          Mouse Genome 430 2.0 Array 1421565_at - -
          Mouse Genome 430 2.0 Array 1421565_PM_at - -
          Mouse Genome 430 2.0 Array 1436634_PM_at - -
          Mouse Genome 430 2.0 Array 1436634_at - -
          Affymetrix Mouse Clariom S Array PSR0900017103.mm.1 NM_001164767.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017103.mm.1 NM_001164767.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017103.mm.1 TC0900002050.mm.1 MTA TC
          Mouse Expression Set 430 1421565_at - -
          Mouse Expression Set 430 1421565_PM_at - -
          Mouse Expression Set 430 1436634_PM_at - -
          Mouse Expression Set 430 1436634_at - -
          Murine Genome U74v2 Set 102081_PM_at - -
          Murine Genome U74v2 Set 102081_at - -
          Mouse Genome 430A 2.0 Array 1421565_at - -
          Mouse Genome 430A 2.0 Array 1421565_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421565_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421565_at - -
          Affymetrix HT Mouse Genome 430B Array 1436634_at - -
          Affymetrix HT Mouse Genome 430B Array 1436634_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 678804 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline