Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00490975_s1
          See other CALM4 GE Assays ›
          Gene Symbol:
          Calm4
          Gene Name:
          calmodulin 4
          Gene Aliases:
          2310037J09Rik, AI649108, DD112, Scarf
          Chromosome Location:
          Chr.13: 3837757 - 3838671 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00490975_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Calm4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020036.4 NP_064420.2 1 394 100
          GenBank mRNA AK003648.1 - 1 395 100
          AK009664.1 - 1 396 100
          AK009956.1 - 1 395 100
          AY293058.1 - 1 377 100
          BC060284.1 - 1 380 30293362 100
          BC145379.1 - 1 350 9053937 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000013.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Calm4

          Panther Classification:

          Molecular Function -

          calmodulin-related

          Gene Ontology Categories:

          Process(es)

          calcium ion binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P576720 BC060284.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P576720 AK009956.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P576720 AY293058.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P576720 NM_020036.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P576720 AK003648.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P576720 BC060284.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P576720 AK009956.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P576720 AY293058.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P576720 NM_020036.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P576720 AK003648.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 390516 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145301 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P576720 BC060284.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P576720 AK009956.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P576720 AY293058.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P576720 NM_020036.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P576720 AK003648.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P576720 BC060284.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P576720 AK009956.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P576720 AY293058.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P576720 NM_020036.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P576720 AK003648.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P460611 BC060284.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P460611 AK009956.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P460611 BC145379.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P460611 AY293058.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P460611 NM_020036.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28790084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28790062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28790058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28790064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28790059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28790090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28790081 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P145301 - -
          Whole Mouse Genome G4122A A_52_P576720 BC060284.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P576720 AK009956.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P576720 AY293058.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P576720 NM_020036.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P576720 AK003648.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P576720 BC060284.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P576720 AK009956.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P576720 AY293058.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P576720 NM_020036.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P576720 AK003648.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P460611 BC060284.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P460611 AK009956.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P460611 BC145379.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P460611 AY293058.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P460611 NM_020036.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116489 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P145301 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10577280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10874679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11019310 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11019318 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11094490 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10403282 - -
          Affymetrix HT MG-430 PM Array Plate 1450633_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10403281 - -
          Mouse Genome 430 2.0 Array 1450633_PM_at - -
          Mouse Genome 430 2.0 Array 1450633_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 80796_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 80796_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300000093.mm.1 NM_020036.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000093.mm.1 NM_020036.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000093.mm.1 TC1300000013.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300000095.mm.1 NM_020036.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000095.mm.1 NM_020036.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000095.mm.1 TC1300000013.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300000094.mm.1 NM_020036.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000094.mm.1 NM_020036.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000094.mm.1 TC1300000013.mm.1 MTA TC
          Mouse Expression Set 430 1450633_PM_at - -
          Mouse Expression Set 430 1450633_at - -
          Murine Genome U74v2 Set 93744_PM_at - -
          Murine Genome U74v2 Set 93744_at - -
          Mouse Genome 430A 2.0 Array 1450633_at - -
          Mouse Genome 430A 2.0 Array 1450633_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450633_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450633_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 909555 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 60026 - -
          Illumina mouse-6 v1.0 expression beadchip 60026 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000060026 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1219326 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1219326 - -
          mouse-6 v1.1 (Illumina) scl080796.1_17-S - -
          Illumina Sentrix mouse-6.v1 bead array 60026 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl080796.1_17-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 60026 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1219326 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1219326 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline