Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00492602_m1
          See other IFI204 GE Assays ›
          Gene Symbol:
          Ifi204
          Gene Name:
          interferon activated gene 204
          Gene Aliases:
          Ifi16, p204
          Chromosome Location:
          Chr.1: 173747293 - 173767068 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00492602_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ifi204
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008329.2 NP_032355.2 6 - 7 1356 71
          GenBank mRNA BC010546.1 - 1334 4018506 71
          BU694586.1 - 2 - 3 278 71
          CT010351.1 - 1248 71
          M31419.1 - 1417 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003888.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ifi204

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          cellular response to interferon-beta
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          positive regulation of osteoblast differentiation
          inner ear development
          regulation of nucleic acid-templated transcription
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          core promoter binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          double-stranded DNA binding
          transcription cofactor activity
          protein binding
          transcription factor binding
          identical protein binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1975560 M31419.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1975560 BC010546.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1975560 BE686930.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1975560 NM_008329.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 472568 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 305237 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 465431 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 809155 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 678110 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 56869 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 63016 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173111 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145095 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2066578 BC010546.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1975560 M31419.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1975560 BC010546.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1975560 BE686930.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1975560 NM_008329.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P408346 M31419.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P408346 BC010546.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P408346 CT010351.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P408346 NM_008329.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P408343 M31419.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P408343 BC010546.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P408343 BE686930.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P408343 CT010351.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P408343 NM_008329.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04359617 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04359571 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173111 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04359601 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04359707 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04359537 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04359634 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04359522 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P145095 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04359498 - -
          Whole Mouse Genome G4122A A_51_P408343 M31419.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408343 BC010546.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408343 BE686930.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408343 CT010351.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408343 NM_008329.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408346 M31419.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408346 BC010546.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408346 CT010351.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408346 NM_008329.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P83479 FJ594741.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P83479 AK087931.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130334 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04359498 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04359634 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04359521 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173111 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04359713 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P145095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839415 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01049043 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839338 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00691517 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839961 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839375 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839328 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839342 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00351015 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00973692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839914 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839346 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839324 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00947639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839958 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839364 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00337094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839950 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839965 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00980900 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839333 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10360389 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360384 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360385 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360386 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360388 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360390 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360383 - -
          Affymetrix HT MG-430 PM Array Plate 1442828_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1419603_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10360382 - -
          Mouse Genome 430 2.0 Array 1419603_PM_at - -
          Mouse Genome 430 2.0 Array 1419603_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15951_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15951_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100029504.mm.1 NM_008329.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029504.mm.1 NM_008329.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029504.mm.1 TC0100003888.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100029508.mm.1 NM_008329.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029508.mm.1 NM_008329.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029508.mm.1 TC0100003888.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100029502.mm.1 NM_008329.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029502.mm.1 NM_008329.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029502.mm.1 TC0100003888.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100029506.mm.1 NM_008329.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029506.mm.1 NM_008329.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029506.mm.1 TC0100003888.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100029507.mm.1 NM_008329.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029507.mm.1 NM_008329.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029507.mm.1 TC0100003888.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100029500.mm.1 NM_008329.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029500.mm.1 NM_008329.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029500.mm.1 TC0100003888.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100029505.mm.1 NM_008329.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029505.mm.1 NM_008329.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029505.mm.1 TC0100003888.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100029501.mm.1 NM_008329.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029501.mm.1 NM_008329.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029501.mm.1 TC0100003888.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100029509.mm.1 NM_008329.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029509.mm.1 NM_008329.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029509.mm.1 TC0100003888.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100029503.mm.1 NM_008329.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029503.mm.1 NM_008329.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029503.mm.1 TC0100003888.mm.1 MTA TC
          Mouse Expression Set 430 1419603_at - -
          Mouse Expression Set 430 1452348_s_at - -
          Mouse Expression Set 430 1419603_PM_at - -
          Mouse Expression Set 430 1452348_PM_s_at - -
          Murine Genome U74v2 Set 98466_PM_r_at - -
          Murine Genome U74v2 Set 94224_PM_s_at - -
          Murine Genome U74v2 Set 98466_r_at - -
          Murine Genome U74v2 Set 161180_r_at - -
          Murine Genome U74v2 Set 98465_PM_f_at - -
          Murine Genome U74v2 Set 94224_s_at - -
          Murine Genome U74v2 Set 161180_PM_r_at - -
          Murine Genome U74v2 Set 98465_f_at - -
          Mouse Genome 430A 2.0 Array 1452348_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1452348_s_at - -
          Mouse Genome 430A 2.0 Array 1419603_at - -
          Mouse Genome 430A 2.0 Array 1419603_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419603_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419603_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 742535 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6520601 - -
          Illumina mouse-6 v1.0 expression beadchip 6520601 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006520601 - -
          mouse-6 v1.1 (Illumina) scl15898.31.1_40-S - -
          Illumina Sentrix mouse-6.v1 bead array 6520601 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl15898.31.1_40-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6520601 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3010089 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3010089 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2623314 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline