Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00494099_m1
          See other PRAP1 GE Assays ›
          Gene Symbol:
          Prap1
          Gene Name:
          proline-rich acidic protein 1
          Gene Aliases:
          Upa
          Chromosome Location:
          Chr.7: 140093396 - 140097203 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00494099_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prap1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009475.2 NP_033501.2 2 - 3 141 90
          GenBank mRNA AA060392.1 - 2 - 3 139 88
          BC051092.1 - 2 - 3 129 6529097 90
          U28486.1 - 2 - 3 140 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002001.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prap1

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P375783 U28486.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P375783 U28486.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 382943 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 953376 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 52932 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141778 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160462 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P375783 U28486.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P375783 U28486.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582207 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160462 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141778 - -
          Whole Mouse Genome G4122A A_51_P375783 U28486.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P375783 U28486.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118643 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160462 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141778 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06065100 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06065093 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06065092 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06065102 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06065099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05966117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05966125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05966128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06179763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05966114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05966116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06028406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05966133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05966107 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10558595 - -
          Affymetrix Mouse Gene 1.0 ST Array 10558591 - -
          Affymetrix Mouse Gene 1.0 ST Array 10558592 - -
          Affymetrix Mouse Gene 1.0 ST Array 10558593 - -
          Affymetrix Mouse Gene 1.0 ST Array 10558594 - -
          Affymetrix HT MG-430 PM Array Plate 1455996_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1419167_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10558590 - -
          Mouse Genome 430 2.0 Array 1419167_at - -
          Mouse Genome 430 2.0 Array 1455996_PM_x_at - -
          Mouse Genome 430 2.0 Array 1455996_x_at - -
          Mouse Genome 430 2.0 Array 1419167_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22264_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22264_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700017580.mm.1 TC0700002001.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700017580.mm.1 NM_009475.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017580.mm.1 NM_009475.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017578.mm.1 TC0700002001.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700017578.mm.1 NM_009475.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017578.mm.1 NM_009475.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017579.mm.1 TC0700002001.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700017579.mm.1 NM_009475.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017579.mm.1 NM_009475.2 Refseq
          Mouse Expression Set 430 1455996_x_at - -
          Mouse Expression Set 430 1419167_PM_at - -
          Mouse Expression Set 430 1455996_PM_x_at - -
          Mouse Expression Set 430 1419167_at - -
          Murine Genome U74v2 Set 99118_PM_at - -
          Murine Genome U74v2 Set 99118_at - -
          Mouse Genome 430A 2.0 Array 1455996_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1419167_at - -
          Mouse Genome 430A 2.0 Array 1419167_PM_at - -
          Mouse Genome 430A 2.0 Array 1455996_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1455996_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1419167_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419167_at - -
          Affymetrix HT Mouse Genome 430A Array 1455996_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 383002 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3830133 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003830133 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1259344 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1259344 - -
          mouse-6 v1.1 (Illumina) scl31918.5.1_81-S - -
          Illumina Sentrix mouse-6.v1 bead array 3830133 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31918.5.1_81-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3830133 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1259344 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1259344 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline