Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00498502_s1
          See other TAS2R105 GE Assays ›
          Gene Symbol:
          Tas2r105
          Gene Name:
          taste receptor, type 2, member 105
          Gene Aliases:
          T2R05, T2R9, T2r5, Tas2r5, mGR05, mt2r5
          Chromosome Location:
          Chr.6: 131686561 - 131687463 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00498502_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tas2r105
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020501.1 NP_065247.1 1 50 88
          GenBank mRNA BC125515.1 - 1 82 40130030 88
          BC125541.1 - 1 82 40130056 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600003275.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tas2r105

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          response to stimulus
          sensory perception of taste
          detection of chemical stimulus involved in sensory perception of taste
          sensory perception of bitter taste
          signal transducer activity
          G-protein coupled receptor activity
          taste receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2150727 NM_020501.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2150727 BC125515.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2150727 BC125541.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100638 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2150727 NM_020501.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2150727 BC125515.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2150727 BC125541.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P177364 NM_020501.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P177364 BC125515.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P177364 BC125541.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24851689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24851693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24851695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24851690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24851680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24851694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24851681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24851684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24851686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24851687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24851678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24851685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24851696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24851677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24851683 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100638 - -
          Whole Mouse Genome G4122A A_51_P177364 NM_020501.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P177364 BC125515.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P177364 BC125541.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120319 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100638 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05689024 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05689027 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05689025 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05689022 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05689012 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05689023 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05689028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24851647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24851680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05689014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24851696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24851687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24851685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24851678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24851693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24851681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24851686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24851682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24851684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05729902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24851691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24851689 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10548613 - -
          Affymetrix HT MG-430 PM Array Plate 1422389_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10548612 - -
          Mouse Genome 430 2.0 Array 1422389_PM_at - -
          Mouse Genome 430 2.0 Array 1422389_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57252_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57252_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600025751.mm.1 TC0600003275.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025751.mm.1 NM_020501.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025751.mm.1 NM_020501.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025752.mm.1 TC0600003275.mm.1 MTA TC
          Mouse Expression Set 430 1422389_PM_at - -
          Mouse Expression Set 430 1422389_at - -
          Mouse Genome 430A 2.0 Array 1422389_at - -
          Mouse Genome 430A 2.0 Array 1422389_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422389_at - -
          Affymetrix HT Mouse Genome 430A Array 1422389_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2450672 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002450672 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2668941 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2668941 - -
          mouse-6 v1.1 (Illumina) scl715.1.1_192-S - -
          Illumina Sentrix mouse-6.v1 bead array 2450672 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl715.1.1_192-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2450672 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2898982 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2898982 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline