Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00499716_m1
          See other TAS1R2 GE Assays ›
          Gene Symbol:
          Tas1r2
          Gene Name:
          taste receptor, type 1, member 2
          Gene Aliases:
          Gpr71, T1r2, TR2
          Chromosome Location:
          Chr.4: 139653538 - 139670280 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00499716_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tas1r2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031873.1 NP_114079.1 4 - 5 1509 83
          GenBank mRNA AF337041.1 - 4 - 5 1509 83
          AY032623.1 - 4 - 5 1478 83
          BC137740.1 - 5 - 6 8860759 83
          BC144879.1 - 5 - 6 9053427 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400004197.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tas1r2

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of chemical stimulus involved in sensory perception of sweet taste
          signal transduction
          G-protein coupled receptor signaling pathway
          positive regulation of cytokinesis
          response to stimulus
          sensory perception of taste
          sensory perception of sweet taste
          signal transducer activity
          G-protein coupled receptor activity
          protein binding
          taste receptor activity
          sweet taste receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23353414 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23353412 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23353413 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P277756 AF337041.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P277756 NM_031873.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P277756 AF337041.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P277756 NM_031873.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 228627 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 352346 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 664802 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 157782 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 160110 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 284929 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 345360 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 834189 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 579484 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 153907 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 127323 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 151714 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 71630 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P277756 AF337041.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P277756 NM_031873.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P277756 AF337041.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P277756 NM_031873.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P277756 AF337041.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P277756 NM_031873.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P277756 AF337041.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P277756 NM_031873.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23353304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23353303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23353328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23353330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23353313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23353318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23353308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23353336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23353316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23353339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23353334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23353307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23353338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23353312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23353311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23353332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23353322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23353324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23353341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23353340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23353333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23353326 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05425753 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01004151 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01004143 - -
          Whole Mouse Genome G4122A A_51_P277756 AF337041.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P277756 NM_031873.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P277756 AF337041.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P277756 NM_031873.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104855 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01004151 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03383714 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03383720 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03865123 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03383716 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23353414 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23353412 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23353413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23353436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353312 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353324 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03819605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04097605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03383717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03368115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353328 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03819608 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353452 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03368116 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03879323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353457 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353446 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03879322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353338 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353342 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04007226 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03383724 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03557882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03608393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03869496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03953215 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03516105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03608400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03865124 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03728509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353454 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03997864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03601633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04007216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04007211 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04007217 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23353347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03217845 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10509815 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509816 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509814 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509817 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509813 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509818 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509812 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509819 - -
          Affymetrix HT MG-430 PM Array Plate 1450313_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10509811 - -
          Mouse Genome 430 2.0 Array 1450313_PM_at - -
          Mouse Genome 430 2.0 Array 1450313_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 83770_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 83770_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400014510.mm.1 NM_031873.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014510.mm.1 NM_031873.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014510.mm.1 TC0400004197.mm.1 MTA TC
          Mouse Expression Set 430 1450313_PM_at - -
          Mouse Expression Set 430 1450313_at - -
          Mouse Genome 430A 2.0 Array 1450313_PM_at - -
          Mouse Genome 430A 2.0 Array 1450313_at - -
          Affymetrix HT Mouse Genome 430A Array 1450313_at - -
          Affymetrix HT Mouse Genome 430A Array 1450313_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 536581 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4850072 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004850072 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2714817 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2714817 - -
          mouse-6 v1.1 (Illumina) scl24769.6.1_288-S - -
          Illumina Sentrix mouse-6.v1 bead array 4850072 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24769.6.1_288-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4850072 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2714817 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2714817 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline