Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00499822_m1
          See other IL25 GE Assays ›
          Gene Symbol:
          Il25
          Gene Name:
          interleukin 25
          Gene Aliases:
          IL-17e, Il17e
          Chromosome Location:
          Chr.14: 54929401 - 54935841 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00499822_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Il25
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080729.3 NP_542767.1 2 - 3 330 82
          GenBank mRNA AF458060.1 - 1 - 2 254 82
          AY034088.1 - 1 - 2 206 82
          BC120568.1 - 2 - 3 329 40129491 82
          BC125524.1 - 2 - 3 329 40130039 82
          BC144962.1 - 2 - 3 329 9053510 82
          BX517868.1 - 1 - 2 139 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400000762.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Il25

          Gene Ontology Categories:

          Function(s) Process(es)

          inflammatory response to antigenic stimulus
          inflammatory response
          response to fungus
          response to nematode
          eosinophil differentiation
          positive regulation of transcription from RNA polymerase II promoter
          cytokine activity
          interleukin-17E receptor binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293015 BC125524.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293015 BC144962.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293015 BC120568.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293015 NM_080729.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293015 AF458060.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293015 BC125524.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293015 BC144962.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293015 BC120568.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293015 NM_080729.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293015 AF458060.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 905378 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 242791 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 59206 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166656 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293015 BC125524.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293015 BC144962.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293015 BC120568.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293015 NM_080729.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293015 AF458060.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293015 BC125524.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293015 BC144962.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293015 BC120568.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293015 NM_080729.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293015 AF458060.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P293015 BC125524.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P293015 BC144962.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P293015 BC120568.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P293015 NM_080729.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P293015 AF458060.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P293015 BC125524.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P293015 BC144962.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P293015 BC120568.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P293015 NM_080729.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P293015 AF458060.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615167 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166656 - -
          Whole Mouse Genome G4122A A_51_P293015 BC125524.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293015 BC144962.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293015 BC120568.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293015 NM_080729.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293015 AF458060.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293015 BC125524.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293015 BC144962.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293015 BC120568.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293015 NM_080729.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293015 AF458060.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122458 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166656 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11230305 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11230321 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11230319 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11230307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11230326 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11796930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11230319 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11230306 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11623028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11230327 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615122 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615154 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10415131 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415130 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415129 - -
          Affymetrix HT MG-430 PM Array Plate 1420740_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10415127 - -
          Mouse Genome 430 2.0 Array 1420740_PM_at - -
          Mouse Genome 430 2.0 Array 1420740_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 140806_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 140806_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1400005936.mm.1 NM_080729.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400005936.mm.1 NM_080729.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400005936.mm.1 TC1400000762.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400005935.mm.1 NM_080729.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400005935.mm.1 NM_080729.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400005935.mm.1 TC1400000762.mm.1 MTA TC
          Mouse Expression Set 430 1420740_PM_at - -
          Mouse Expression Set 430 1420740_at - -
          Murine Genome U74v2 Set 132535_at - -
          Murine Genome U74v2 Set 132535_PM_at - -
          Mouse Genome 430A 2.0 Array 1420740_PM_at - -
          Mouse Genome 430A 2.0 Array 1420740_at - -
          Affymetrix HT Mouse Genome 430A Array 1420740_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420740_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 60041 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000060041 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2695495 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2695495 - -
          mouse-6 v1.1 (Illumina) scl46295.2.1_28-S - -
          Illumina Sentrix mouse-6.v1 bead array 60041 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46295.2.1_28-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 60041 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2695495 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2695495 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline