Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00499866_m1
          See other SLC7A12 GE Assays ›
          Gene Symbol:
          Slc7a12
          Gene Name:
          solute carrier family 7 (cationic amino acid transporter, y+ system), member 12
          Gene Aliases:
          Asc-2, XAT1
          Chromosome Location:
          Chr.3: 14480691 - 14505819 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00499866_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc7a12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080852.2 NP_543128.1 1 - 2 774 88
          GenBank mRNA AB055005.1 - 1 - 2 779 88
          AJ417663.1 - 1 - 2 736 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000089.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Transporter Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc7a12

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          amino acid transmembrane transport
          amino acid transport
          L-alpha-amino acid transmembrane transport
          amino acid transmembrane transporter activity
          L-amino acid transmembrane transporter activity
          antiporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2061984 AB055005.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2061984 AJ417663.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2061984 NM_080852.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 730089 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 277192 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 255358 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 527247 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 687858 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 385315 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 526593 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 786622 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 5293 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 174831 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128758 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164160 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2061984 AB055005.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2061984 AJ417663.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2061984 NM_080852.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369823 AB055005.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369823 AJ417663.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369823 NM_080852.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21957075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21957078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21957069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21957070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21957076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21957083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21957072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21957085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21957084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21957087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21957082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21957068 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P128758 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04831545 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00588695 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P164160 - -
          Whole Mouse Genome G4122A A_51_P369823 AB055005.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369823 AJ417663.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369823 NM_080852.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122896 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125460 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P128758 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04831539 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P164160 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02631817 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02631819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21957103 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21957082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21957087 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21957072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21957083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21957080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21957076 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02550786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21957084 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21957078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02674351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21957075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21957085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02550784 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10490868 - -
          Affymetrix Mouse Gene 1.0 ST Array 10490871 - -
          Affymetrix Mouse Gene 1.0 ST Array 10490870 - -
          Affymetrix Mouse Gene 1.0 ST Array 10490869 - -
          Affymetrix HT MG-430 PM Array Plate 1419579_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10490867 - -
          Mouse Genome 430 2.0 Array 1419579_at - -
          Mouse Genome 430 2.0 Array 1419579_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 140918_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 140918_at - -
          Affymetrix Mouse Clariom S Array PSR0300000333.mm.1 NM_080852.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300000333.mm.1 NM_080852.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300000333.mm.1 TC0300000089.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300000330.mm.1 NM_080852.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300000330.mm.1 NM_080852.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300000330.mm.1 TC0300000089.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300000334.mm.1 NM_080852.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300000334.mm.1 NM_080852.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300000334.mm.1 TC0300000089.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300000335.mm.1 NM_080852.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300000335.mm.1 NM_080852.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300000335.mm.1 TC0300000089.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300000332.mm.1 NM_080852.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300000332.mm.1 NM_080852.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300000332.mm.1 TC0300000089.mm.1 MTA TC
          Mouse Expression Set 430 1419579_at - -
          Mouse Expression Set 430 1419579_PM_at - -
          Mouse Genome 430A 2.0 Array 1419579_PM_at - -
          Mouse Genome 430A 2.0 Array 1419579_at - -
          Affymetrix HT Mouse Genome 430A Array 1419579_at - -
          Affymetrix HT Mouse Genome 430A Array 1419579_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 362321 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1090402 - -
          Illumina mouse-6 v1.0 expression beadchip 1090402 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001090402 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2633726 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2633726 - -
          mouse-6 v1.1 (Illumina) scl23384.4.1_41-S - -
          Illumina Sentrix mouse-6.v1 bead array 1090402 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl23384.4.1_41-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1090402 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2633726 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2633726 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline