Search
Search

Runx2| Gene Symbol: Runx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145920.2 | NP_001139392.1 | 2 - 3 | 1576 | 129 | |
| NM_001146038.2 | NP_001139510.1 | 4 - 5 | 841 | 129 | ||
| NM_001271627.1 | NP_001258556.1 | 3 - 4 | 1005 | 129 | ||
| NM_001271630.1 | NP_001258559.1 | 2 - 3 | 1104 | 129 | ||
| NM_009820.5 | NP_033950.2 | 4 - 5 | 808 | 129 | ||
| NR_073392.1 | 3 - 4 | 592 | 129 | |||
| NR_073425.1 | 2 - 3 | 1104 | 129 | |||
| GenBank mRNA | AF005936.1 | - | 3 - 4 | 959 | 129 | |
| AF010284.1 | - | 4 - 5 | 808 | 129 | ||
| AF053954.1 | - | 3 - 4 | 422 | 129 | ||
| AF053955.1 | - | 2 - 3 | 256 | 129 | ||
| AK134925.1 | - | 2 - 3 | 362 | 129 | ||
| AK158925.1 | - | 2 - 3 | 1104 | 129 | ||
| D14636.1 | - | 2 - 3 | 1574 | 129 | ||
| D14637.1 | - | 2 - 3 | 1574 | 129 | ||
| DQ458792.1 | - | 3 - 4 | 593 | 129 | ||
Set Membership: |
|