Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00503141_m1
          See other ERP29 GE Assays ›
          Gene Symbol:
          Erp29
          Gene Name:
          endoplasmic reticulum protein 29
          Gene Aliases:
          1200015M03Rik, 2810446M09Rik, AW209030, Erp28, Erp31, PDI-Db
          Chromosome Location:
          Chr.5: 121444753 - 121452474 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00503141_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Erp29
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026129.2 NP_080405.1 2 - 3 421 75
          GenBank mRNA AK004795.1 - 2 - 3 324 75
          AK009881.1 - 2 - 3 327 75
          AK154539.1 - 2 - 3 317 75
          AK165702.1 - 321 75
          AK168112.1 - 2 - 3 341 75
          BC017125.1 - 2 - 3 421 4481563 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003188.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Erp29

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          positive regulation of protein phosphorylation
          ER to Golgi vesicle-mediated transport
          protein secretion
          positive regulation of gene expression
          negative regulation of gene expression
          negative regulation of protein secretion
          regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
          protein homodimerization activity
          chaperone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24044667 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24044663 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24044670 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24044669 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24044665 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24044668 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24044664 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049383 BC017125.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049383 AK154539.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049383 AK165702.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049383 AK168112.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049383 NM_026129.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2094362 BC017125.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2094362 AK154539.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2094362 AK165702.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2094362 AK168112.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2094362 NM_026129.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 113417 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 745793 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 269318 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 38802 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 927629 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 11678 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 968712 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 41040 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 25196 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157295 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P110698 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2094362 BC017125.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2094362 AK154539.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2094362 AK165702.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2094362 AK168112.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2094362 NM_026129.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049383 BC017125.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049383 AK154539.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049383 AK165702.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049383 AK168112.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049383 NM_026129.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044708 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044700 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24044685 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01205645 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157295 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P110698 - -
          Whole Mouse Genome G4122A A_51_P133670 BC017125.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P133670 AK154539.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P133670 AK165702.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P133670 AK168112.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P133670 NM_026129.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121222 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157295 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P110698 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04225787 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04672770 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04225785 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04158392 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04153819 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24044665 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24044670 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24044668 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24044664 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24044669 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24044667 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24044663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24044634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04960124 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04736256 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04225785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04744781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04575390 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044677 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04191098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04173055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04960127 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044612 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044609 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04453017 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04783689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044610 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044613 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044624 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044663 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044670 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04672769 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04783690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04960123 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24044620 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10533319 - -
          Affymetrix Mouse Gene 1.0 ST Array 10533318 - -
          Affymetrix Mouse Gene 1.0 ST Array 10533317 - -
          Affymetrix HT MG-430 PM Array Plate 1453634_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10533316 - -
          Mouse Genome 430 2.0 Array 1453634_a_at - -
          Mouse Genome 430 2.0 Array 1453634_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67397_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67397_at - -
          Affymetrix Mouse Clariom S Array PSR0500028377.mm.1 TC0500003188.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500028377.mm.1 NM_026129.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500028377.mm.1 NM_026129.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500028378.mm.1 TC0500003188.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500028378.mm.1 NM_026129.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500028378.mm.1 NM_026129.2 Refseq
          Murine Genome U74v2 Set 161755_PM_at - -
          Murine Genome U74v2 Set 98076_at - -
          Murine Genome U74v2 Set 98076_PM_at - -
          Murine Genome U74v2 Set 161755_at - -
          Affymetrix HT Mouse Genome 430A Array 1453634_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1453634_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 917423 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4050441 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004050441 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1239185 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1239185 - -
          mouse-6 v1.1 (Illumina) scl067397.1_307-S - -
          Illumina Sentrix mouse-6.v1 bead array 4050441 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl067397.1_307-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4050441 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1239185 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1239185 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl067397.1_307-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline