Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00504709_m1
          See other GLMN GE Assays ›
          Gene Symbol:
          Glmn
          Gene Name:
          glomulin, FKBP associated protein
          Gene Aliases:
          9330160J16Rik, AW227515, Fap48, Fap68
          Chromosome Location:
          Chr.5: 107548967 - 107597888 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00504709_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Glmn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001161738.1 NP_001155210.1 8 - 9 1044 77
          NM_001161739.1 NP_001155211.1 8 - 9 1058 77
          NM_133248.2 NP_573511.2 8 - 9 1058 77
          GenBank mRNA AJ566083.1 - 7 - 8 926 77
          AK034160.1 - 8 - 9 1045 77
          AK165661.1 - 8 - 9 1029 77
          AK169045.1 - 8 - 9 1030 77
          AK172659.1 - 8 - 9 996 77
          BC003446.1 - 8 - 9 994 2651614 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002927.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Glmn

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          vasculogenesis
          neural tube closure
          cell surface receptor signaling pathway
          negative regulation of cell proliferation
          negative regulation of protein ubiquitination
          regulation of proteasomal ubiquitin-dependent protein catabolic process
          regulation of gene expression, epigenetic
          negative regulation of T cell proliferation
          positive regulation of phosphorylation
          muscle cell differentiation
          negative regulation of catalytic activity
          positive regulation of interleukin-2 biosynthetic process
          positive regulation of cytokine secretion
          circulatory system development
          receptor binding
          hepatocyte growth factor receptor binding
          protein binding
          ubiquitin protein ligase binding
          ubiquitin-protein transferase inhibitor activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500025869.mm.1 TC0500002927.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500025869.mm.1 NM_001161738.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500025869.mm.1 NM_001161738.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500025869.mm.1 NM_001161739.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500025869.mm.1 NM_001161739.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500025869.mm.1 NM_133248.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500025869.mm.1 NM_133248.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 733467 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0