Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00504792_m1
          See other PRSS8 GE Assays ›
          Gene Symbol:
          Prss8
          Gene Name:
          protease, serine 8 (prostasin)
          Gene Aliases:
          2410039E18Rik, AI313909, C79772, CAP1, fr, mCAP1
          Chromosome Location:
          Chr.7: 127925717 - 127930142 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00504792_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prss8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133351.3 NP_579929.1 2 - 3 338 71
          GenBank mRNA AB038244.1 - 2 - 3 319 71
          AF188613.1 - 2 - 3 252 71
          AF378085.2 - 2 - 3 329 71
          AK010640.1 - 2 - 3 221 71
          AK078696.1 - 2 - 3 290 71
          AY335911.1 - 3 - 4 186 71
          BC003851.1 - 2 - 3 312 3600399 71
          CB600036.1 - 2 - 3 341 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004346.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prss8

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          hair follicle development
          proteolysis
          positive regulation of sodium ion transport
          transepithelial transport
          serine-type endopeptidase activity
          peptidase activity
          serine-type peptidase activity
          hydrolase activity
          sodium channel regulator activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700036521.mm.1 NM_133351.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036521.mm.1 NM_133351.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036521.mm.1 TC0700004346.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036523.mm.1 NM_133351.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036523.mm.1 NM_133351.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036523.mm.1 TC0700004346.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036522.mm.1 NM_133351.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036522.mm.1 NM_133351.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036522.mm.1 TC0700004346.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036518.mm.1 NM_133351.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036518.mm.1 NM_133351.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036518.mm.1 TC0700004346.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036519.mm.1 NM_133351.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036519.mm.1 NM_133351.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036519.mm.1 TC0700004346.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 579377 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline