Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00505672_m1
          See other ZBTB48 GE Assays ›
          Gene Symbol:
          Zbtb48
          Gene Name:
          zinc finger and BTB domain containing 48
          Gene Aliases:
          0610011D15Rik, AI327031, Hkr3
          Chromosome Location:
          Chr.4: 152019773 - 152027671 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00505672_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zbtb48
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133879.2 NP_598640.2 3 - 4 1040 105
          GenBank mRNA AK027912.1 - 2 - 3 1409 105
          AM161454.1 - 3 - 4 988 105
          BC005515.1 - 3 - 4 1051 3493611 105
          CX202255.1 - 1 - 2 217 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400004088.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zbtb48

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          biological_process
          nucleic acid binding
          DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23416102 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23416100 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23416103 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P370341 BC005515.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P370341 BC005515.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 404733 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 99534 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 66267 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584599 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 130850 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P370341 BC005515.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P370341 BC005515.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P370341 BC005515.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P370341 BC005515.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416103 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416100 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416093 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416095 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416102 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416099 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05453860 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01020549 - -
          Whole Mouse Genome G4122A A_51_P370341 BC005515.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P370341 BC005515.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133834 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03349795 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23416103 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23416100 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23416102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23415997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23415963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23415969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23415974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23415970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23415991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23415965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23415984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23415982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23415995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23415977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23415979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23415975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23415976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23415967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23415986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23415973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23415971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23415989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23415987 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416087 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416050 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03349814 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03349823 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03318048 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416046 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03318005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416048 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416097 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03349792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03250906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03318034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03318033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03349802 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03349813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416102 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03289087 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03318031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416084 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416069 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03318014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416081 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416064 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416079 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10518865 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518858 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518861 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518867 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518866 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518868 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518860 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518859 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518862 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518863 - -
          Affymetrix HT MG-430 PM Array Plate 1418200_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10518857 - -
          Mouse Genome 430 2.0 Array 1418200_PM_at - -
          Mouse Genome 430 2.0 Array 1418200_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 100090_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 100090_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400034018.mm.1 TC0400004088.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400034018.mm.1 NM_133879.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400034018.mm.1 NM_133879.2 Refseq
          Mouse Expression Set 430 1418200_PM_at - -
          Mouse Expression Set 430 1418200_at - -
          Murine Genome U74v2 Set 103765_at - -
          Murine Genome U74v2 Set 103765_PM_at - -
          Murine Genome U74v2 Set 168748_r_at - -
          Murine Genome U74v2 Set 168748_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1418200_PM_at - -
          Mouse Genome 430A 2.0 Array 1418200_at - -
          Affymetrix HT Mouse Genome 430A Array 1418200_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418200_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 887225 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5550458 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005550458 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2726532 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2726532 - -
          mouse-6 v1.1 (Illumina) scl23469.10.1_15-S - -
          Illumina Sentrix mouse-6.v1 bead array 5550458 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl23469.10.1_15-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5550458 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline