Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00506761_m1
          See other YIPF2 GE Assays ›
          Gene Symbol:
          Yipf2
          Gene Name:
          Yip1 domain family, member 2
          Gene Aliases:
          1300010K09Rik
          Chromosome Location:
          Chr.9: 21588682 - 21592831 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00506761_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Yipf2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001205157.1 NP_001192086.1 5 - 6 487 64
          NM_001205158.1 NP_001192087.1 5 - 6 477 64
          NM_138303.2 NP_612176.1 4 - 5 625 64
          NR_038062.1 4 - 5 426 64
          GenBank mRNA AK004959.1 - 4 - 5 411 64
          AK033705.1 - 5 - 6 436 64
          AK133631.1 - 5 - 6 447 64
          BC002282.1 - 4 - 5 540 3497525 64
          BC003289.1 - 5 - 6 418 3583087 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001879.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Yipf2

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26347945 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26347951 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26347947 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170474 BC002282.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170474 BC003289.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170474 AK004959.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170474 AK133631.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170474 NM_001205158.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 275277 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 134195 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P178956 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147342 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100058 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P122574 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170474 BC002282.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170474 BC003289.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170474 AK004959.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170474 AK133631.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170474 NM_001205158.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347947 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P122574 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P147342 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P178956 - -
          Whole Mouse Genome G4122A A_52_P338162 BC002282.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338162 BC003289.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338162 AK004959.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338162 AK133631.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338162 NM_001205158.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P534150 BC002282.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P534150 BC003289.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P534150 AK004959.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P534150 AK033705.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P534150 AK133631.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P305731 BC002282.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P305731 BC003289.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P305731 AK004959.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P305731 AK133631.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P305731 NM_001205158.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108995 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115182 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P122574 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P147342 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P178956 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07836186 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26347945 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26347947 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26347951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07836167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07489239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347950 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10591554 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591548 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591551 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591549 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591555 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591547 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591546 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591550 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591553 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591552 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591545 - -
          Affymetrix HT MG-430 PM Array Plate 1438415_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1420668_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10583676 - -
          Affymetrix Mouse Gene 1.1 ST Array 10591544 - -
          Mouse Genome 430 2.0 Array 1420668_PM_a_at - -
          Mouse Genome 430 2.0 Array 1438415_PM_s_at - -
          Mouse Genome 430 2.0 Array 1438415_s_at - -
          Mouse Genome 430 2.0 Array 1420668_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74766_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74766_at - -
          Affymetrix Mouse Clariom S Array PSR0900015779.mm.1 NM_001205157.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015779.mm.1 NM_001205157.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015779.mm.1 NM_001205158.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015779.mm.1 NM_001205158.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015779.mm.1 NM_138303.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015779.mm.1 NM_138303.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015779.mm.1 NR_038062.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015779.mm.1 NR_038062.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015779.mm.1 TC0900001879.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900015780.mm.1 NM_001205157.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015780.mm.1 NM_001205157.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015780.mm.1 NM_001205158.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015780.mm.1 NM_001205158.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015780.mm.1 NM_138303.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015780.mm.1 NM_138303.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015780.mm.1 NR_038062.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015780.mm.1 NR_038062.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015780.mm.1 TC0900001879.mm.1 MTA TC
          Mouse Expression Set 430 1420668_a_at - -
          Mouse Expression Set 430 1438415_PM_s_at - -
          Mouse Expression Set 430 1420668_PM_a_at - -
          Mouse Expression Set 430 1438415_s_at - -
          Mouse Genome 430A 2.0 Array 1420668_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1438415_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1420668_a_at - -
          Mouse Genome 430A 2.0 Array 1438415_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1438415_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1420668_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1420668_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1438415_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 746069 - -
          ABI Mouse Genome Survey Microarray 931268 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3390181 - -
          Illumina mouse-6 v1.0 expression beadchip 2470168 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003390181 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002470168 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1239874 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2691566 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2657849 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1239874 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2657849 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2691566 - -
          mouse-6 v1.1 (Illumina) scl36137.8_30-S - -
          mouse-6 v1.1 (Illumina) scl0003559.1_19-S - -
          Illumina Sentrix mouse-6.v1 bead array 2470168 - -
          Illumina Sentrix mouse-6.v1 bead array 3390181 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36137.8_30-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0003559.1_19-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2470168 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3390181 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2691566 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2691566 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl36137.8_30-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0