Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00506799_m1
          See other NELFE GE Assays ›
          Gene Symbol:
          Nelfe
          Gene Name:
          negative elongation factor complex member E, Rdbp
          Gene Aliases:
          D17H6S45, NELF-E, RD, Rdbp
          Chromosome Location:
          Chr.17: 34850391 - 34856372 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00506799_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nelfe
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001045863.1 NP_001039328.1 4 - 5 494 74
          NM_001045864.1 NP_001039329.1 4 - 5 580 74
          NM_138580.2 NP_613046.2 4 - 5 368 74
          GenBank mRNA AK011663.1 - 4 - 5 352 74
          AK012389.1 - 4 - 5 385 74
          AK131947.1 - 4 - 5 352 74
          AK147011.1 - 4 - 5 476 74
          BC034867.1 - 4 - 5 320 1495547 74
          CN525174.1 - 4 - 5 323 74
          DV662511.1 - 1 - 2 113 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000647.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nelfe

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          negative regulation of transcription elongation from RNA polymerase II promoter
          negative regulation of mRNA polyadenylation
          nucleotide binding
          nucleic acid binding
          RNA binding
          protein binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31156113 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P110270 AK131947.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P110270 AK147011.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P110270 BC034867.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P110270 NM_001045864.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P110270 NM_138580.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P110270 AK131947.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P110270 AK147011.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P110270 BC034867.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P110270 NM_001045864.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P110270 NM_138580.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 874466 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 430164 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 202176 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 704652 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 18926 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152599 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P110270 AK131947.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P110270 AK147011.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P110270 BC034867.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P110270 NM_001045864.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P110270 NM_138580.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P110270 AK131947.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P110270 AK147011.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P110270 BC034867.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P110270 NM_001045864.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P110270 NM_138580.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P110270 AK131947.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P110270 AK147011.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P110270 BC034867.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P110270 NM_001045864.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P110270 NM_138580.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P110270 AK131947.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P110270 AK147011.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P110270 BC034867.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P110270 NM_001045864.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P110270 NM_138580.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156119 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152599 - -
          Whole Mouse Genome G4122A A_52_P110270 AK131947.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110270 AK147011.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110270 BC034867.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110270 NM_001045864.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110270 NM_138580.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110270 AK131947.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110270 AK147011.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110270 BC034867.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110270 NM_001045864.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110270 NM_138580.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131572 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152599 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13241295 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13241290 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13241292 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13241286 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31156113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13090886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13552804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13546115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13241289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13552805 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13404645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13546114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13241286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13546110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13546116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13546106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13546112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156122 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156143 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156115 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10444515 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444511 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444509 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444508 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444506 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444514 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444518 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444519 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444520 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444512 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444510 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444516 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444517 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444513 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444507 - -
          Affymetrix HT MG-430 PM Array Plate 1450705_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10444505 - -
          Mouse Genome 430 2.0 Array 1450705_PM_at - -
          Mouse Genome 430 2.0 Array 1450705_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 27632_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 27632_at - -
          Affymetrix Mouse Clariom S Array PSR1700006404.mm.1 NM_001045863.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006404.mm.1 NM_001045863.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006404.mm.1 NM_001045864.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006404.mm.1 NM_001045864.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006404.mm.1 NM_138580.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006404.mm.1 NM_138580.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006404.mm.1 TC1700000647.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700006405.mm.1 NM_001045863.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006405.mm.1 NM_001045863.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006405.mm.1 NM_001045864.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006405.mm.1 NM_001045864.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006405.mm.1 NM_138580.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006405.mm.1 NM_138580.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006405.mm.1 TC1700000647.mm.1 MTA TC
          Mouse Expression Set 430 1450705_at - -
          Mouse Expression Set 430 1450705_PM_at - -
          Murine Genome U74v2 Set 92610_at - -
          Murine Genome U74v2 Set 92610_PM_at - -
          Mouse Genome 430A 2.0 Array 1450705_PM_at - -
          Mouse Genome 430A 2.0 Array 1450705_at - -
          Affymetrix HT Mouse Genome 430A Array 1450705_at - -
          Affymetrix HT Mouse Genome 430A Array 1450705_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 764339 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6590044 - -
          MouseWG-6 v2 Expression BeadChip 105720497 - -
          Illumina mouse-6 v1.0 expression beadchip 105720497 - -
          Illumina mouse-6 v1.0 expression beadchip 6590044 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006590044 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1233513 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1253999 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1233513 - -
          mouse-6 v1.1 (Illumina) scl50808.8.1_29-S - -
          mouse-6 v1.1 (Illumina) scl0027632.2_240-S - -
          Illumina Sentrix mouse-6.v1 bead array 105720497 - -
          Illumina Sentrix mouse-6.v1 bead array 6590044 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0027632.2_240-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105720497 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6590044 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3094608 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1233513 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3094608 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1233513 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1253999 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0027632.2_240-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline