Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00509876_m1
          See other PTGES3L GE Assays ›
          Gene Symbol:
          Ptges3l
          Gene Name:
          prostaglandin E synthase 3 (cytosolic)-like
          Gene Aliases:
          1110069E20Rik, 1700113I22Rik
          Chromosome Location:
          Chr.11: 101418801 - 101425333 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00509876_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptges3l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026865.2 NP_081141.1 4 - 5 1000 63
          GenBank mRNA AK004415.1 - 4 - 5 318 63
          AK007198.1 - 4 - 5 999 63
          BC057970.1 - 4 - 5 306 63
          BC141198.1 - 4 - 5 407 9055810 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003804.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ptges3l

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073809 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073805 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073803 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073806 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073804 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073808 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073807 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P126563 AK007198.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2144745 AK007198.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2144745 BC141198.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2144745 NM_026865.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 289795 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 393924 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 362692 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 806324 AK090132.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 806324 AK137564.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 806324 AK147263.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 806324 AK147627.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 806324 AK147642.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 806324 AK147676.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 806324 NM_007872.4 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 806324 NM_153743.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 940354 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 126382 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171417 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P126563 AK007198.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2144745 AK007198.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2144745 BC141198.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2144745 NM_026865.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073820 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073832 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073814 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073807 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073826 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07466602 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171417 - -
          Whole Mouse Genome G4122A A_52_P480096 AK007198.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P480096 BC057970.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P480096 AK004415.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P480096 NM_026865.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126563 AK007198.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171417 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09561704 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09561706 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09561700 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09561697 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09503637 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073803 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073805 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073807 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073809 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073808 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073804 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09634949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08993001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09561700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09561703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09561692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09385614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073808 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09361905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09361902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073832 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10391446 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391445 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391450 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391453 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391452 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391448 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391451 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391449 - -
          Affymetrix HT MG-430 PM Array Plate 1432508_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1432073_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1454256_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10391444 - -
          Mouse Genome 430 2.0 Array 1454256_s_at - -
          Mouse Genome 430 2.0 Array 1432073_PM_at - -
          Mouse Genome 430 2.0 Array 1432508_at - -
          Mouse Genome 430 2.0 Array 1454256_PM_s_at - -
          Mouse Genome 430 2.0 Array 1432508_PM_at - -
          Mouse Genome 430 2.0 Array 1432073_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73635_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73635_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100035230.mm.1 NM_026865.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035230.mm.1 NM_026865.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035230.mm.1 TC1100003804.mm.1 MTA TC
          Mouse Expression Set 430 1424006_PM_at - -
          Mouse Expression Set 430 1424006_at - -
          Mouse Expression Set 430 1454256_s_at - -
          Mouse Expression Set 430 1432073_at - -
          Mouse Expression Set 430 1454256_PM_s_at - -
          Mouse Expression Set 430 1432508_PM_at - -
          Mouse Expression Set 430 1432508_at - -
          Mouse Expression Set 430 1432073_PM_at - -
          Murine Genome U74v2 Set 171578_PM_at - -
          Murine Genome U74v2 Set 171578_at - -
          Mouse Genome 430A 2.0 Array 1424006_PM_at - -
          Mouse Genome 430A 2.0 Array 1424006_at - -
          Affymetrix HT Mouse Genome 430B Array 1432508_at - -
          Affymetrix HT Mouse Genome 430B Array 1432508_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2320520 - -
          Illumina mouse-6 v1.0 expression beadchip 2320520 - -
          Illumina mouse-6 v1.0 expression beadchip 3440647 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003440647 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002320520 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2662036 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2596569 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2662036 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2596569 - -
          mouse-6 v1.1 (Illumina) scl0001450.1_42-S - -
          mouse-6 v1.1 (Illumina) scl39533.9.1_23-S - -
          Illumina Sentrix mouse-6.v1 bead array 2320520 - -
          Illumina Sentrix mouse-6.v1 bead array 3440647 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001450.1_42-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl39533.9.1_23-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3440647 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2320520 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2662036 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2596569 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2662036 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2596569 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline