Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00511056_m1
          See other TMCO6 GE Assays ›
          Gene Symbol:
          Tmco6
          Gene Name:
          transmembrane and coiled-coil domains 6
          Gene Aliases:
          2410015B03Rik
          Chromosome Location:
          Chr.18: 36735018 - 36742400 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00511056_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmco6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028036.3 NP_082312.3 2 - 3 301 88
          GenBank mRNA AK010487.1 - 2 - 3 228 88
          AK046207.1 - 2 - 3 332 88
          BC005637.1 - 2 - 3 303 3156734 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000342.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK010487.1,AK046207.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmco6

          Gene Ontology Categories:

          Function(s) Process(es)

          protein import into nucleus
          biological_process
          protein transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625518 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625517 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625516 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292368 BC005637.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292368 BF468256.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292368 NM_028036.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292368 AK010487.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292368 BC005637.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292368 BF468256.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292368 NM_028036.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292368 AK010487.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 766500 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 96011 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 302569 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 669401 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 422339 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 123786 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170165 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292368 BC005637.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292368 BF468256.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292368 NM_028036.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292368 AK010487.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292368 BC005637.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292368 BF468256.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292368 NM_028036.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292368 AK010487.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625558 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P170165 - -
          Whole Mouse Genome G4122A A_51_P292368 BC005637.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P292368 BF468256.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P292368 NM_028036.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P292368 AK010487.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P292368 BC005637.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P292368 BF468256.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P292368 NM_028036.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P292368 AK010487.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124626 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109225 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P170165 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13887757 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13887763 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625516 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625518 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13666518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13777943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14039649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13942419 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13969936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13887764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13777941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13666516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13777940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14039650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14117686 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10454957 - -
          Affymetrix Mouse Gene 1.0 ST Array 10454959 - -
          Affymetrix Mouse Gene 1.0 ST Array 10454955 - -
          Affymetrix Mouse Gene 1.0 ST Array 10454962 - -
          Affymetrix Mouse Gene 1.0 ST Array 10454961 - -
          Affymetrix Mouse Gene 1.0 ST Array 10454965 - -
          Affymetrix Mouse Gene 1.0 ST Array 10454956 - -
          Affymetrix Mouse Gene 1.0 ST Array 10454954 - -
          Affymetrix Mouse Gene 1.0 ST Array 10454964 - -
          Affymetrix Mouse Gene 1.0 ST Array 10454958 - -
          Affymetrix Mouse Gene 1.0 ST Array 10454963 - -
          Affymetrix Mouse Gene 1.0 ST Array 10454960 - -
          Affymetrix HT MG-430 PM Array Plate 1459587_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1451692_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10454953 - -
          Mouse Genome 430 2.0 Array 1459587_PM_at - -
          Mouse Genome 430 2.0 Array 1459587_at - -
          Mouse Genome 430 2.0 Array 1451692_at - -
          Mouse Genome 430 2.0 Array 1451692_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71983_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71983_at - -
          Affymetrix Mouse Clariom S Array PSR1800002606.mm.1 NM_028036.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002606.mm.1 NM_028036.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002606.mm.1 TC1800000342.mm.1 MTA TC
          Mouse Expression Set 430 1451692_PM_at - -
          Mouse Expression Set 430 1451692_at - -
          Mouse Expression Set 430 1459587_at - -
          Mouse Expression Set 430 1459587_PM_at - -
          Mouse Genome 430A 2.0 Array 1451692_at - -
          Mouse Genome 430A 2.0 Array 1451692_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451692_at - -
          Affymetrix HT Mouse Genome 430A Array 1451692_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1459587_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1459587_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 420175 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2850348 - -
          MouseWG-6 v2 Expression BeadChip 1990463 - -
          Illumina mouse-6 v1.0 expression beadchip 2680487 - -
          Illumina mouse-6 v1.0 expression beadchip 2850348 - -
          Illumina mouse-6 v1.0 expression beadchip 1990463 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002680487 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002850348 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001990463 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2621853 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232025 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2620037 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2620037 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2621853 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232025 - -
          mouse-6 v1.1 (Illumina) scl52082.9.1_8-S - -
          mouse-6 v1.1 (Illumina) scl0002151.1_1-S - -
          mouse-6 v1.1 (Illumina) scl0002248.1_32-S - -
          Illumina Sentrix mouse-6.v1 bead array 2850348 - -
          Illumina Sentrix mouse-6.v1 bead array 2680487 - -
          Illumina Sentrix mouse-6.v1 bead array 1990463 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0002151.1_1-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0002248.1_32-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52082.9.1_8-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2680487 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2850348 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1990463 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2621853 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2620037 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1232025 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1232025 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2620037 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2621853 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline