Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00513411_m1
          See other GINS3 GE Assays ›
          Gene Symbol:
          Gins3
          Gene Name:
          GINS complex subunit 3 (Psf3 homolog)
          Gene Aliases:
          2700085M18Rik, AI616142
          Chromosome Location:
          Chr.8: 95633559 - 95645059 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00513411_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gins3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030198.3 NP_084474.1 1 - 2 228 119
          GenBank mRNA AK019225.1 - 1 - 2 229 119
          AK146574.1 - 1 - 2 259 119
          AK150602.1 - 1 - 2 260 119
          AK151243.1 - 1 - 2 260 119
          AK152572.1 - 1 - 2 260 119
          AK160606.1 - 1 - 2 229 119
          BC039794.1 - 1 - 2 225 5121680 119
          BC064746.1 - 1 - 2 217 30095310 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001142.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gins3

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA replication
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26084323 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26084325 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2025090 BC064746.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2025090 AK152572.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2025090 AK160606.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2025090 AK177395.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2025090 BC039794.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 800781 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 696623 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731373 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 789187 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 573011 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 675044 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 212024 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 646897 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 8811 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176102 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151293 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2025090 BC064746.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2025090 AK152572.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2025090 AK160606.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2025090 AK177395.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2025090 BC039794.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26084316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26084313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26084334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26084304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26084330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26084307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26084324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26084298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26084321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26084295 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26084332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26084310 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26084314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26084336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26084326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26084301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26084327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26084319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26084296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26084308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26084337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26084325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26084305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26084328 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P176102 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06605852 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151293 - -
          Whole Mouse Genome G4122A A_51_P130957 BC064746.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P130957 AK160606.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P130957 AK146574.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P130957 NM_030198.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P130957 AK019225.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101645 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151293 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P176102 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06938421 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06938423 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06938435 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06938438 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06938436 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06938428 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26084323 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26084325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26084343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07426336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07026664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06944610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06938428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06762393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07134135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07218219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07026661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06960077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06938423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06938438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07002402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07115980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07433469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26084299 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10574381 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574383 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574382 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574379 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574380 - -
          Affymetrix HT MG-430 PM Array Plate 1453050_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1429149_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10574378 - -
          Mouse Genome 430 2.0 Array 1440421_PM_at - -
          Mouse Genome 430 2.0 Array 1429149_PM_at - -
          Mouse Genome 430 2.0 Array 1453050_PM_at - -
          Mouse Genome 430 2.0 Array 1429149_at - -
          Mouse Genome 430 2.0 Array 1440421_at - -
          Mouse Genome 430 2.0 Array 1453050_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78833_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78833_at - -
          Affymetrix Mouse Clariom S Array PSR0800008980.mm.1 NM_030198.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800008980.mm.1 NM_030198.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800008980.mm.1 TC0800001142.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800008981.mm.1 NM_030198.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800008981.mm.1 NM_030198.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800008981.mm.1 TC0800001142.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800008982.mm.1 NM_030198.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800008982.mm.1 NM_030198.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800008982.mm.1 TC0800001142.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800008978.mm.1 NM_030198.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800008978.mm.1 NM_030198.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800008978.mm.1 TC0800001142.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800008979.mm.1 NM_030198.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800008979.mm.1 NM_030198.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800008979.mm.1 TC0800001142.mm.1 MTA TC
          Mouse Expression Set 430 1429149_at - -
          Mouse Expression Set 430 1429149_PM_at - -
          Mouse Expression Set 430 1453050_PM_at - -
          Mouse Expression Set 430 1453050_at - -
          Affymetrix HT Mouse Genome 430B Array 1453050_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453050_at - -
          Affymetrix HT Mouse Genome 430B Array 1429149_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429149_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 540172 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 610576 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000610576 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2589960 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2589960 - -
          mouse-6 v1.1 (Illumina) scl078833.3_67-S - -
          Illumina Sentrix mouse-6.v1 bead array 610576 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl078833.3_67-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 610576 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2589960 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2589960 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline