Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00517966_m1
          See other CCDC59 GE Assays ›
          Gene Symbol:
          Ccdc59
          Gene Name:
          coiled-coil domain containing 59
          Gene Aliases:
          2300004H16Rik, D10Ertd718e
          Chromosome Location:
          Chr.10: 105841479 - 105847510 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00517966_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccdc59
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025602.3 NP_079878.1 1 - 2 180 131
          GenBank mRNA AK009028.1 - 1 - 2 547 131
          AK012917.1 - 1 - 2 163 131
          AK019970.1 - 1 - 2 181 131
          AK047009.1 - 1 - 2 585 131
          AK135550.1 - 1 - 2 205 131
          BC013721.1 - 1 - 2 563 131
          BC027401.1 - 1 - 2 165 4948072 131

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000001255.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccdc59

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          biological_process
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27418184 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27418183 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27418182 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P129795 AK135550.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P129795 BC027401.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P129795 NM_025602.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P129795 AK012917.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P129795 AK019970.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 407833 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 532617 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 702156 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 47943 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174746 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100076 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106375 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P129795 AK135550.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P129795 BC027401.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P129795 NM_025602.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P129795 AK012917.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P129795 AK019970.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27418211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27418195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27418192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27418214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27418198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27418210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27418199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27418220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27418216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27418188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27418202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27418193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27418209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27418212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27418185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27418206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27418219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27418184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27418197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27418191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27418187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27418207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27418204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27418223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27418196 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106375 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174746 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100076 - -
          Whole Mouse Genome G4122A A_52_P557995 AK047009.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P557995 AK135550.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P557995 BC013721.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P557995 BC027401.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P557995 NM_025602.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130360 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129795 AK135550.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129795 BC027401.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129795 NM_025602.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129795 AK012917.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129795 AK019970.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174746 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100076 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106375 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08846709 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08846708 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08846722 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27418183 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27418182 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27418184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27418185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27418215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27418199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27418238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08328310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27418232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27418223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27418207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27418239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27418214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27418225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27418182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27418193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27418195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27418237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27418234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27418236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27418192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08846723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27418187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27418185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27418219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08328303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27418211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08328313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27418184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27418231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27418229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08328311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08465997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27418202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27418201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27418204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27418216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08846711 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10366187 - -
          Affymetrix Mouse Gene 1.0 ST Array 10366189 - -
          Affymetrix Mouse Gene 1.0 ST Array 10366188 - -
          Affymetrix Mouse Gene 1.0 ST Array 10366191 - -
          Affymetrix Mouse Gene 1.0 ST Array 10366190 - -
          Affymetrix HT MG-430 PM Array Plate 1449446_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10366186 - -
          Mouse Genome 430 2.0 Array 1449446_at - -
          Mouse Genome 430 2.0 Array 1449446_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 52713_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 52713_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000009441.mm.1 NM_025602.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000009441.mm.1 NM_025602.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000009441.mm.1 TC1000001255.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000009439.mm.1 NM_025602.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000009439.mm.1 NM_025602.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000009439.mm.1 TC1000001255.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000009440.mm.1 NM_025602.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000009440.mm.1 NM_025602.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000009440.mm.1 TC1000001255.mm.1 MTA TC
          Mouse Expression Set 430 1449446_PM_at - -
          Mouse Expression Set 430 1449446_at - -
          Murine Genome U74v2 Set 167437_r_at - -
          Murine Genome U74v2 Set 167437_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1449446_PM_at - -
          Mouse Genome 430A 2.0 Array 1449446_at - -
          Affymetrix HT Mouse Genome 430A Array 1449446_at - -
          Affymetrix HT Mouse Genome 430A Array 1449446_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 380181 - -
          Illumina mouse-6 v1.0 expression beadchip 380181 - -
          Illumina mouse-6 v1.0 expression beadchip 4210273 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000380181 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004210273 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2712584 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1256220 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256220 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2712584 - -
          mouse-6 v1.1 (Illumina) scl052713.2_45-S - -
          mouse-6 v1.1 (Illumina) scl0052713.1_274-S - -
          Illumina Sentrix mouse-6.v1 bead array 4210273 - -
          Illumina Sentrix mouse-6.v1 bead array 380181 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl052713.2_45-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0052713.1_274-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4210273 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 380181 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2712584 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2877436 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1256220 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2877436 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1256220 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2712584 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl052713.2_45-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0