Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00521959_m1
          See other LY6D GE Assays ›
          Gene Symbol:
          Ly6d
          Gene Name:
          lymphocyte antigen 6 complex, locus D
          Gene Aliases:
          Ly-61, Ly61, Thb
          Chromosome Location:
          Chr.15: 74762056 - 74763567 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00521959_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ly6d
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010742.1 NP_034872.1 1 - 2 57 65
          GenBank mRNA BC025135.1 - 1 - 2 52 4914126 65
          X63782.1 - 1 - 2 57 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001718.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ly6d

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343517 NM_010742.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343517 BC025135.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343517 X63782.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343517 NM_010742.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343517 BC025135.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343517 X63782.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 209527 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 593920 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108878 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179486 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118753 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343517 NM_010742.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343517 BC025135.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343517 X63782.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343517 NM_010742.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343517 BC025135.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343517 X63782.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P343517 NM_010742.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P343517 BC025135.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P343517 X63782.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P343517 NM_010742.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P343517 BC025135.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P343517 X63782.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339470 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339475 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339471 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339482 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339466 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339505 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P108878 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179486 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118753 - -
          Whole Mouse Genome G4122A A_51_P343517 NM_010742.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343517 BC025135.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343517 X63782.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343517 NM_010742.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343517 BC025135.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343517 X63782.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106499 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P108878 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118753 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179486 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11855915 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12095897 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12095891 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12095903 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12095901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339483 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11855905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12414574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12095894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339505 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339502 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339467 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12414578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12286073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339491 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12120624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339465 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11906594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339469 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12414585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11855918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11918538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12095901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12120623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12095903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12365124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12365125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12286072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11918536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339466 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339479 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12095891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12120630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339536 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10429522 - -
          Affymetrix Mouse Gene 1.0 ST Array 10429521 - -
          Affymetrix Mouse Gene 1.0 ST Array 10429523 - -
          Affymetrix HT MG-430 PM Array Plate 1416930_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10429520 - -
          Mouse Genome 430 2.0 Array 1416930_PM_at - -
          Mouse Genome 430 2.0 Array 1416930_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17068_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17068_at - -
          Affymetrix Mouse Clariom S Array PSR1500012973.mm.1 NM_010742.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012973.mm.1 NM_010742.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012973.mm.1 TC1500001718.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500012972.mm.1 NM_010742.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012972.mm.1 NM_010742.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012972.mm.1 TC1500001718.mm.1 MTA TC
          Mouse Expression Set 430 1416930_PM_at - -
          Mouse Expression Set 430 1416930_at - -
          Murine Genome U74v2 Set 162216_PM_i_at - -
          Murine Genome U74v2 Set 160553_PM_at - -
          Murine Genome U74v2 Set 162216_i_at - -
          Murine Genome U74v2 Set 160553_at - -
          Mouse Genome 430A 2.0 Array 1416930_at - -
          Mouse Genome 430A 2.0 Array 1416930_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416930_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416930_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 577604 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4050010 - -
          Illumina mouse-6 v1.0 expression beadchip 4050010 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004050010 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1257839 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1257839 - -
          mouse-6 v1.1 (Illumina) scl47080.2.4_26-S - -
          Illumina Sentrix mouse-6.v1 bead array 4050010 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl47080.2.4_26-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4050010 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2803921 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2803920 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2803921 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2803920 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline