Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00523063_m1
          See other NDUFS8 GE Assays ›
          Gene Symbol:
          Ndufs8
          Gene Name:
          NADH dehydrogenase (ubiquinone) Fe-S protein 8
          Gene Aliases:
          BC021616, CI-23kD, TYKY
          Chromosome Location:
          Chr.19: 3908863 - 3912774 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00523063_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ndufs8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001271443.1 NP_001258372.1 5 - 6 441 58
          NM_001271444.1 NP_001258373.1 5 - 6 681 58
          NM_144870.5 NP_659119.2 5 - 6 684 58
          GenBank mRNA AK135069.1 - 5 - 6 442 58
          AK149674.1 - 5 - 6 642 58
          AK153182.1 - 5 - 6 628 58
          AK153900.1 - 5 - 6 418 58
          AK164901.1 - 5 - 6 418 58
          AK167907.1 - 5 - 6 419 58
          BC021616.1 - 5 - 6 402 5132866 58
          BC086766.1 - 5 - 6 395 6820786 58
          BP775598.1 - 2 - 3 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000889.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ndufs8

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          response to oxidative stress
          mitochondrial respiratory chain complex I assembly
          oxidation-reduction process
          NADH dehydrogenase activity
          NADH dehydrogenase (ubiquinone) activity
          oxidoreductase activity
          oxidoreductase activity, acting on NAD(P)H
          metal ion binding
          iron-sulfur cluster binding
          4 iron, 4 sulfur cluster binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P138829 BC021616.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 869300 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 667590 AK014768.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 667590 AK031808.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 667590 AK033623.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 667590 AK038156.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 667590 AK039879.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 667590 AK042500.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 667590 AK050983.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 667590 BC033266.1 genbank_mrna_mouse
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131420 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P138829 BC021616.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P247441 BC021616.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P247441 BC086766.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P247441 NM_144870.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P247441 AK135069.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P247441 AK149674.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31919979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31919966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31919985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31919993 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31919972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31919996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31919982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31919976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31919987 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31919973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31919980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31919969 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31919986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31919964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31919995 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31919967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31919983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31919990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31919981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31919974 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31919977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31919994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31919998 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131420 - -
          Whole Mouse Genome G4122A A_51_P247441 BC021616.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P247441 BC086766.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P247441 NM_144870.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P247441 AK135069.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P247441 AK149674.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138829 BC021616.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131420 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14487466 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14487461 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14487471 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14487452 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14487456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31920068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31919991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31919983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14487452 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31919969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31919990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31919966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14291096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31919976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14487456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31919967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14218515 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14218511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31919987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31919980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31919974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31919985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31919972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31919996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31919993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14487466 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14218503 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31919964 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10464558 - -
          Affymetrix Mouse Gene 1.0 ST Array 10464554 - -
          Affymetrix Mouse Gene 1.0 ST Array 10464559 - -
          Affymetrix Mouse Gene 1.0 ST Array 10464555 - -
          Affymetrix Mouse Gene 1.0 ST Array 10464556 - -
          Affymetrix Mouse Gene 1.0 ST Array 10464557 - -
          Affymetrix Mouse Gene 1.0 ST Array 10464553 - -
          Affymetrix Mouse Gene 1.0 ST Array 10464552 - -
          Affymetrix HT MG-430 PM Array Plate 1423908_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1434213_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1423907_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1455283_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1434212_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1434579_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10464551 - -
          Mouse Genome 430 2.0 Array 1423908_PM_at - -
          Mouse Genome 430 2.0 Array 1423907_a_at - -
          Mouse Genome 430 2.0 Array 1434213_x_at - -
          Mouse Genome 430 2.0 Array 1434212_PM_at - -
          Mouse Genome 430 2.0 Array 1423907_PM_a_at - -
          Mouse Genome 430 2.0 Array 1434212_at - -
          Mouse Genome 430 2.0 Array 1434579_PM_x_at - -
          Mouse Genome 430 2.0 Array 1455283_x_at - -
          Mouse Genome 430 2.0 Array 1434213_PM_x_at - -
          Mouse Genome 430 2.0 Array 1455283_PM_x_at - -
          Mouse Genome 430 2.0 Array 1434579_x_at - -
          Mouse Genome 430 2.0 Array 1423908_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 225887_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 225887_at - -
          Affymetrix Mouse Clariom S Array PSR1900007696.mm.1 NM_001271443.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007696.mm.1 NM_001271444.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007696.mm.1 NM_144870.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007696.mm.1 TC1900000889.mm.1 MTA TC
          Mouse Expression Set 430 1434213_x_at - -
          Mouse Expression Set 430 1455283_PM_x_at - -
          Mouse Expression Set 430 1423907_a_at - -
          Mouse Expression Set 430 1423908_at - -
          Mouse Expression Set 430 1434212_at - -
          Mouse Expression Set 430 1434212_PM_at - -
          Mouse Expression Set 430 1434579_x_at - -
          Mouse Expression Set 430 1434579_PM_x_at - -
          Mouse Expression Set 430 1423907_PM_a_at - -
          Mouse Expression Set 430 1455283_x_at - -
          Mouse Expression Set 430 1434213_PM_x_at - -
          Mouse Expression Set 430 1423908_PM_at - -
          Murine Genome U74v2 Set 165405_at - -
          Murine Genome U74v2 Set 165405_PM_at - -
          Mouse Genome 430A 2.0 Array 1434579_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1434213_x_at - -
          Mouse Genome 430A 2.0 Array 1434212_PM_at - -
          Mouse Genome 430A 2.0 Array 1423908_PM_at - -
          Mouse Genome 430A 2.0 Array 1434212_at - -
          Mouse Genome 430A 2.0 Array 1434213_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1434579_x_at - -
          Mouse Genome 430A 2.0 Array 1423907_a_at - -
          Mouse Genome 430A 2.0 Array 1455283_x_at - -
          Mouse Genome 430A 2.0 Array 1423908_at - -
          Mouse Genome 430A 2.0 Array 1455283_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1423907_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1455283_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1423907_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1434213_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1434212_at - -
          Affymetrix HT Mouse Genome 430A Array 1455283_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1434579_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1423908_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1434213_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1423907_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1423908_at - -
          Affymetrix HT Mouse Genome 430A Array 1434212_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1434579_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 781482 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4150100 - -
          Illumina mouse-6 v1.0 expression beadchip 4150100 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004150100 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2711734 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2711734 - -
          mouse-6 v1.1 (Illumina) scl53435.8.1_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 4150100 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl53435.8.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4150100 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2711734 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2711734 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl53435.8.1_0-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline