Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00523810_m1
          See other ZFP189 GE Assays ›
          Gene Symbol:
          Zfp189
          Gene Name:
          zinc finger protein 189
          Gene Aliases:
          BB131811, C430015I23Rik
          Chromosome Location:
          Chr.4: 49521176 - 49531558 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00523810_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp189
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289901.1 NP_001276830.1 3 - 4 336 77
          NM_145547.3 NP_663522.2 2 - 3 507 77
          GenBank mRNA AK049475.1 - 2 - 3 416 77
          AK051265.1 - 2 - 3 466 77
          BC021326.1 - 2 - 3 469 5065942 77
          BC087833.1 - 2 - 3 373 5708354 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400000552.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp189

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of defense response to virus by host
          regulation of transcription, DNA-templated
          mitophagy in response to mitochondrial depolarization
          transcription factor activity, sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22905512 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22905511 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2065646 BC021326.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2065646 BC087833.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2065646 AK049475.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2065646 AK051265.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2065646 NM_145547.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 847032 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584189 AK035437.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 234827 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 746604 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 132897 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 594206 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 328625 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 68519 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149530 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159365 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2065646 BC021326.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2065646 BC087833.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2065646 AK049475.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2065646 AK051265.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2065646 NM_145547.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P358792 BC087833.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P358792 AK049475.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P358792 AK051265.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P358792 NM_145547.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905528 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149530 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05231881 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159365 - -
          Whole Mouse Genome G4122A A_51_P358792 BC087833.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P358792 AK049475.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P358792 AK051265.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P358792 NM_145547.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132848 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149530 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159365 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03676855 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03988108 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03676845 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03754562 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22905511 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22905512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03298474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03988108 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905549 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03324976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905564 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03662587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905581 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03884416 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03869386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905582 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905513 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03883683 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03884428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905531 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03754562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03662588 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03295285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905552 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04000983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905541 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905580 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10504924 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504923 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504921 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504919 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504920 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504925 - -
          Affymetrix HT MG-430 PM Array Plate 1425287_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10504918 - -
          Mouse Genome 430 2.0 Array 1425287_PM_at - -
          Mouse Genome 430 2.0 Array 1425287_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 230162_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 230162_at - -
          Affymetrix Mouse Clariom S Array PSR0400004043.mm.1 NM_001289901.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004043.mm.1 NM_145547.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004043.mm.1 TC0400000552.mm.1 MTA TC
          Mouse Expression Set 430 1425287_at - -
          Mouse Expression Set 430 1425287_PM_at - -
          Mouse Genome 430A 2.0 Array 1425287_PM_at - -
          Mouse Genome 430A 2.0 Array 1425287_at - -
          Affymetrix HT Mouse Genome 430A Array 1425287_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425287_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 642268 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4730154 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004730154 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2479585 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2479585 - -
          mouse-6 v1.1 (Illumina) scl25444.3_226-S - -
          Illumina Sentrix mouse-6.v1 bead array 4730154 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl25444.3_226-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4730154 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2479585 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2479585 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0