Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00526176_s1
          See other OLFR374 GE Assays ›
          Gene Symbol:
          Olfr374
          Gene Name:
          olfactory receptor 374
          Gene Aliases:
          MOR130-1
          Chromosome Location:
          Chr.8: 72107040 - 72110509 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00526176_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr374
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146338.2 NP_666450.2 2 683 122
          GenBank mRNA BC145662.1 - 1 627 40130965 122

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000812.mm.1
          This assay targets the same location as RTA transcript cluster(s) TC1600000239.rn

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr374

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2069609 NM_146338.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2069609 BC145662.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 48787 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176732 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2069609 NM_146338.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2069609 BC145662.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P374533 NM_146338.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959708 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959705 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959712 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959690 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P176732 - -
          Whole Mouse Genome G4122A A_51_P374533 NM_146338.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136054 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P176732 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06787899 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06811106 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06787894 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06787898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07381701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06787894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07118944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06888009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07378538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06787898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06929556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06787889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07378540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06787890 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10572746 - -
          Affymetrix Mouse Gene 1.1 ST Array 10572745 - -
          Affymetrix Mouse Clariom S Array PSR0800005963.mm.1 NM_146338.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005963.mm.1 NM_146338.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005963.mm.1 TC0800000812.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800005965.mm.1 NM_146338.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005965.mm.1 NM_146338.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005965.mm.1 TC0800000812.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800005964.mm.1 TC0800000812.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 332854 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3800131 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003800131 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2670007 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2670007 - -
          mouse-6 v1.1 (Illumina) scl0258335.1_24-S - -
          Illumina Sentrix mouse-6.v1 bead array 3800131 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258335.1_24-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3800131 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2670007 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2670007 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0