Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00526767_s1
          See other OLFR843 GE Assays ›
          Gene Symbol:
          Olfr843
          Gene Name:
          olfactory receptor 843
          Gene Aliases:
          MOR155-2
          Chromosome Location:
          Chr.9: 19248357 - 19249463 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00526767_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr843
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146567.2 NP_666778.2 1 920 119
          GenBank mRNA BC138386.1 - 1 920 8861406 119
          BC138387.1 - 1 920 8861407 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001815.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr843

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2057851 BC138386.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2057851 NM_146567.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2057851 BC138387.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 835084 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117119 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139809 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126208 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2057851 BC138386.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2057851 NM_146567.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2057851 BC138387.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P175549 BC138386.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P175549 NM_146567.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P175549 BC138387.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339495 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139809 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126208 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P117119 - -
          Whole Mouse Genome G4122A A_51_P175549 BC138386.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P175549 NM_146567.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P175549 BC138387.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100434 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139809 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P117119 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126208 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07938313 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07938315 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07938314 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07938299 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07938308 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07824970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339506 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10591189 - -
          Affymetrix Mouse Gene 1.1 ST Array 10591188 - -
          Affymetrix Mouse Clariom S Array PSR0900015228.mm.1 NM_146567.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015228.mm.1 NM_146567.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015228.mm.1 TC0900001815.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 823006 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4010139 - -
          Illumina mouse-6 v1.0 expression beadchip 4010139 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004010139 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1220060 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1220060 - -
          mouse-6 v1.1 (Illumina) scl36180.1.1_19-S - -
          Illumina Sentrix mouse-6.v1 bead array 4010139 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36180.1.1_19-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4010139 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2591659 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2591659 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline