Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00527021_s1
          See other OLFR697 GE Assays ›
          Gene Symbol:
          Olfr697
          Gene Name:
          olfactory receptor 697
          Gene Aliases:
          MOR283-5
          Chromosome Location:
          Chr.7: 106740905 - 106741956 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00527021_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr697
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146599.2 NP_666810.1 1 487 92
          GenBank mRNA BC138389.1 - 1 487 8861409 92
          BC145999.1 - 1 487 40131302 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004016.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr697

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P243187 NM_146599.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P243187 BC138389.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P243187 BC145999.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P243187 NM_146599.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P243187 BC138389.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P243187 BC145999.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102270 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P243187 NM_146599.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P243187 BC138389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P243187 BC145999.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P243187 NM_146599.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P243187 BC138389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P243187 BC145999.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P243187 NM_146599.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P243187 BC138389.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P243187 BC145999.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P243187 NM_146599.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P243187 BC138389.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P243187 BC145999.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396512 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102270 - -
          Whole Mouse Genome G4122A A_51_P243187 NM_146599.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P243187 BC138389.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P243187 BC145999.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P243187 NM_146599.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P243187 BC138389.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P243187 BC145999.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120443 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102270 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06422351 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06422338 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06482812 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06482807 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06482800 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06422349 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06482814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05999335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06482816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05950073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06658787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06422354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06422341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06252607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396488 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566596 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566595 - -
          Affymetrix Mouse Clariom S Array PSR0700033445.mm.1 NM_146599.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033445.mm.1 NM_146599.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033445.mm.1 TC0700004016.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 319643 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3190300 - -
          Illumina mouse-6 v1.0 expression beadchip 3190300 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003190300 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1223349 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223349 - -
          mouse-6 v1.1 (Illumina) scl30862.1.1_59-S - -
          Illumina Sentrix mouse-6.v1 bead array 3190300 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl30862.1.1_59-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3190300 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1223349 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1223349 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0