Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00528861_s1
          See other OLFR211 GE Assays ›
          Gene Symbol:
          Olfr211
          Gene Name:
          olfactory receptor 211
          Gene Aliases:
          MOR253-9
          Chromosome Location:
          Chr.6: 116489117 - 116494540 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00528861_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr211
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146912.1 NP_667123.1 1 548 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001302.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr211

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163383 NM_146912.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123306 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163383 NM_146912.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P357299 NM_146912.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24776899 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123306 - -
          Whole Mouse Genome G4122A A_51_P357299 NM_146912.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123306 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05415861 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05346578 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05415882 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05415883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24776905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05415870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05775268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05510914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05510921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05415863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05415872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05415866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05775276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05510911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24776935 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10541062 - -
          Affymetrix Mouse Gene 1.1 ST Array 10541061 - -
          Affymetrix Mouse Clariom S Array PSR0600010359.mm.1 TC0600001302.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600010359.mm.1 NM_146912.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600010359.mm.1 NM_146912.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 678230 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4200075 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004200075 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2740062 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2740062 - -
          mouse-6 v1.1 (Illumina) scl1503.1.1_259-S - -
          Illumina Sentrix mouse-6.v1 bead array 4200075 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl1503.1.1_259-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4200075 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2740062 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2740062 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0