Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00546522_g1
          See other PGP GE Assays ›
          Gene Symbol:
          Pgp
          Gene Name:
          phosphoglycolate phosphatase
          Gene Aliases:
          1700012G19Rik, AI481330, AUM
          Chromosome Location:
          Chr.17: 24470473 - 24471602 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00546522_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pgp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025954.3 NP_080230.2 1 - 2 677 76
          GenBank mRNA AK005906.1 - 1 - 2 349 76
          AK010094.1 - 1 - 2 74 76
          BC040100.1 - 1 - 2 677 4456284 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000339.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pgp

          Panther Classification:

          Molecular Function -

          phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          metabolic process
          dephosphorylation
          peptidyl-tyrosine dephosphorylation
          magnesium ion binding
          phosphoprotein phosphatase activity
          protein tyrosine phosphatase activity
          phosphoglycolate phosphatase activity
          hydrolase activity
          phosphatase activity
          metal ion binding
          nucleotide phosphatase activity, acting on free nucleotides

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31097498 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31097494 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31097503 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31097496 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31097500 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31097501 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31097497 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31097499 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466964 AK013578.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466964 BC040100.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466964 BC083113.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466964 NM_025954.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466964 AK005906.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466964 AK013578.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466964 BC040100.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466964 BC083113.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466964 NM_025954.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466964 AK005906.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136854 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466964 AK013578.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466964 BC040100.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466964 BC083113.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466964 NM_025954.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466964 AK005906.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466964 AK013578.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466964 BC040100.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466964 BC083113.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466964 NM_025954.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466964 AK005906.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31097501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31097507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31097483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31097500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31097485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31097486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31097506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31097475 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31097494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31097495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31097503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31097481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31097511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31097491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31097476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31097510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31097490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31097488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31097480 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31097499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31097505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31097504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31097496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31097508 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136854 - -
          Whole Mouse Genome G4122A A_52_P96772 AK013578.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P96772 AK012010.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466964 AK013578.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466964 BC040100.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466964 BC083113.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466964 NM_025954.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466964 AK005906.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466964 AK013578.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466964 BC040100.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466964 BC083113.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466964 NM_025954.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466964 AK005906.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136854 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13074412 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13074407 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13074413 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31097500 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31097497 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31097496 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31097499 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31097503 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31097494 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31097501 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31097498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31097471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13092870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31097469 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13293565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31097489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31097499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31097475 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13293607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31097483 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31097507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31097485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31097476 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31097508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31097481 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13404740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31097474 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31097494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31097490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31097486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13120558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31097491 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31097480 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31097503 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13293578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31097488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13562328 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31097496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31097510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31097504 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442456 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442455 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442457 - -
          Affymetrix HT MG-430 PM Array Plate 1452919_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1428788_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10442454 - -
          Mouse Genome 430 2.0 Array 1452919_PM_a_at - -
          Mouse Genome 430 2.0 Array 1428788_at - -
          Mouse Genome 430 2.0 Array 1452919_a_at - -
          Mouse Genome 430 2.0 Array 1428788_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67078_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67078_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700002535.mm.1 NM_025954.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002535.mm.1 NM_025954.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002535.mm.1 TC1700000339.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002532.mm.1 NM_025954.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002532.mm.1 NM_025954.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002532.mm.1 TC1700000339.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002533.mm.1 NM_025954.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002533.mm.1 NM_025954.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002533.mm.1 TC1700000339.mm.1 MTA TC
          Mouse Expression Set 430 1452919_a_at - -
          Mouse Expression Set 430 1428788_at - -
          Mouse Expression Set 430 1452919_PM_a_at - -
          Mouse Expression Set 430 1428788_PM_at - -
          Murine Genome U74v2 Set 98890_PM_at - -
          Murine Genome U74v2 Set 98890_at - -
          Mouse Genome 430A 2.0 Array 1428788_at - -
          Mouse Genome 430A 2.0 Array 1452919_a_at - -
          Mouse Genome 430A 2.0 Array 1428788_PM_at - -
          Mouse Genome 430A 2.0 Array 1452919_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1452919_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1428788_at - -
          Affymetrix HT Mouse Genome 430A Array 1428788_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452919_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 702956 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 940059 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000940059 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2727318 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2727318 - -
          mouse-6 v1.1 (Illumina) scl067078.2_14-S - -
          Illumina Sentrix mouse-6.v1 bead array 940059 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl067078.2_14-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 940059 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2727318 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2727318 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl067078.2_14-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline