Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00551926_s1
          See other OLFR959 GE Assays ›
          Gene Symbol:
          Olfr959
          Gene Name:
          olfactory receptor 959
          Gene Aliases:
          M34, MOR224-3, ORL583
          Chromosome Location:
          Chr.9: 39572322 - 39573257 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00551926_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr959
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146508.1 NP_666719.1 1 544 78
          GenBank mRNA BC120617.1 - 1 601 40129540 78
          BC120619.1 - 1 601 40129542 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002082.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr959

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466731 BC120619.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466731 NM_146508.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466731 BC120617.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466731 BC120619.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466731 NM_146508.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466731 BC120617.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P172506 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466731 BC120619.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466731 NM_146508.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466731 BC120617.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466731 BC120619.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466731 NM_146508.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466731 BC120617.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P466731 BC120619.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P466731 NM_146508.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P466731 BC120617.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P466731 BC120619.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P466731 NM_146508.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P466731 BC120617.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439113 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439095 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439103 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439099 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439109 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439104 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439106 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439116 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439100 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439098 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P172506 - -
          Whole Mouse Genome G4122A A_51_P466731 BC120619.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466731 NM_146508.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466731 BC120617.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466731 BC120619.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466731 NM_146508.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466731 BC120617.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115125 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P172506 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07505186 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07505181 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07505170 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07505158 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07505180 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07505167 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07505169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07505168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07621162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07874387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07505183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07505167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07874388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07874383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439062 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10592419 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592417 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592418 - -
          Affymetrix Mouse Gene 1.1 ST Array 10592416 - -
          Affymetrix Mouse Clariom S Array PSR0900017212.mm.1 NM_146508.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017212.mm.1 NM_146508.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017212.mm.1 TC0900002082.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900017213.mm.1 TC0900002082.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900017211.mm.1 TC0900002082.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 566210 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6900131 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006900131 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2614582 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2614582 - -
          mouse-6 v1.1 (Illumina) scl6616.1.1_205-S - -
          Illumina Sentrix mouse-6.v1 bead array 6900131 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl6616.1.1_205-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6900131 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2614582 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2614582 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline