Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00613733_m1
          See other 4930544M13RIK GE Assays ›
          Gene Symbol:
          4930544M13Rik
          Gene Name:
          RIKEN cDNA 4930544M13 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 114607228 - 114697822 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00613733_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930544M13Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK076988.1 - 3 - 4 641 84
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA NM_175218.1 - 641 Chr. - - [-] on Build 84

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 4930544M13Rik

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P321244 AK076988.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P321244 BB020030.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P321244 AK076988.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P321244 BB020030.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2260914 AK016040.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 409775 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 468787 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 854635 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731233 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 962581 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 628416 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 311848 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 298884 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 672751 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 108402 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 331104 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 85967 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 405290 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 328841 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 54766 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 122503 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154578 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113378 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347358 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347356 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347346 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29347318 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121389 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154578 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08005073 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08005013 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08004992 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02782987 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08004917 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08004962 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08005041 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113378 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08004975 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08005045 - -
          Whole Mouse Genome G4122A A_52_P321244 AK076988.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P321244 BB020030.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P321244 AK076988.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P321244 BB020030.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129105 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154578 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02782987 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121389 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08004917 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347371 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347431 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347370 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347356 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10439295 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347373 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10712983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347350 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10653554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10646071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347436 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347346 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347429 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347437 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10614492 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347383 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347353 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11062786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347405 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10932585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347357 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347439 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10513250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347368 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10856925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347358 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347401 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347505 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347395 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347386 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10513243 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10589952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347503 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347393 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10457229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10513240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10513237 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10999179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347388 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347403 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347390 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347381 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10768851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347515 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347398 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347374 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10932586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10736061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10999169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10531068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347577 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1431483_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1445818_PM_at - -
          Mouse Genome 430 2.0 Array 1431483_at - -
          Mouse Genome 430 2.0 Array 1445818_PM_at - -
          Mouse Genome 430 2.0 Array 1431483_PM_at - -
          Mouse Genome 430 2.0 Array 1445818_at - -
          Mouse Expression Set 430 1431483_PM_at - -
          Mouse Expression Set 430 1431483_at - -
          Mouse Expression Set 430 1445818_PM_at - -
          Mouse Expression Set 430 1445818_at - -
          Murine Genome U74v2 Set 170617_PM_at - -
          Murine Genome U74v2 Set 170617_at - -
          Affymetrix HT Mouse Genome 430B Array 1445818_at - -
          Affymetrix HT Mouse Genome 430B Array 1431483_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431483_at - -
          Affymetrix HT Mouse Genome 430B Array 1445818_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 397819 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 101500433 - -
          Illumina mouse-6 v1.0 expression beadchip 4730441 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004730441 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2680694 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2476291 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2680694 - -
          mouse-6 v1.1 (Illumina) scl0075161.2_93-S - -
          mouse-6 v1.1 (Illumina) scl44344.8.1_104-S - -
          Illumina Sentrix mouse-6.v1 bead array 4730441 - -
          Illumina Sentrix mouse-6.v1 bead array 101500433 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0075161.2_93-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4730441 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101500433 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2476291 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2680694 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline