Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00614773_m1
          See other CATSPERD GE Assays ›
          Gene Symbol:
          Catsperd
          Gene Name:
          catsper channel auxiliary subunit delta
          Gene Aliases:
          4921529N20Rik, 4933402B14Rik, 619718, AW045609, Gm6095, Tmem146
          Chromosome Location:
          Chr.17: 56626896 - 56664460 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00614773_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Catsperd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175350.3 NP_780559.2 15 - 16 1463 91
          GenBank mRNA AK077082.1 - 6 - 7 524 91
          BC115864.1 - 4 - 5 416 91
          BC116633.1 - 5 - 6 530 40056655 91
          HQ441159.1 - 15 - 16 1455 91
          HQ441160.1 - 12 - 13 1351 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001013.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Catsperd

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          spermatogenesis
          cell differentiation
          sperm motility
          sperm capacitation
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2083009 NM_175350.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170064 HQ441159.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170064 HQ441160.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170064 NM_175350.3 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P172782 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127944 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2083009 NM_175350.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170064 HQ441159.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170064 HQ441160.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170064 NM_175350.3 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127944 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08812073 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P172782 - -
          Whole Mouse Genome G4122A A_51_P401153 HQ441159.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401153 HQ441160.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401153 BC116633.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401153 NM_175350.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401153 AK077082.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107672 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128534 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127944 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P172782 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257464 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13322681 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13428924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257466 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13274472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257495 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13274469 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13165273 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13165274 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13120364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13159067 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257465 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13098900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13309964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257503 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257469 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13089723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13227293 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13359816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13159068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257483 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257461 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257478 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257473 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13089726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13142834 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10446143 - -
          Affymetrix Mouse Gene 1.0 ST Array 10446140 - -
          Affymetrix Mouse Gene 1.0 ST Array 10446141 - -
          Affymetrix Mouse Gene 1.0 ST Array 10446138 - -
          Affymetrix Mouse Gene 1.0 ST Array 10446147 - -
          Affymetrix Mouse Gene 1.0 ST Array 10446146 - -
          Affymetrix Mouse Gene 1.0 ST Array 10446142 - -
          Affymetrix Mouse Gene 1.0 ST Array 10446145 - -
          Affymetrix Mouse Gene 1.0 ST Array 10446137 - -
          Affymetrix Mouse Gene 1.0 ST Array 10446144 - -
          Affymetrix Mouse Gene 1.0 ST Array 10446139 - -
          Affymetrix Mouse Gene 1.0 ST Array 10446148 - -
          Affymetrix HT MG-430 PM Array Plate 1434726_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10446136 - -
          Mouse Genome 430 2.0 Array 1434726_PM_at - -
          Mouse Genome 430 2.0 Array 1434726_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 106757_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 106757_at - -
          Affymetrix Mouse Clariom S Array PSR1700009742.mm.1 NM_175350.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009742.mm.1 NM_175350.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009742.mm.1 TC1700001013.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700009743.mm.1 NM_175350.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009743.mm.1 NM_175350.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009743.mm.1 TC1700001013.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700009741.mm.1 NM_175350.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009741.mm.1 NM_175350.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009741.mm.1 TC1700001013.mm.1 MTA TC
          Mouse Expression Set 430 1434726_at - -
          Mouse Expression Set 430 1434726_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434726_at - -
          Affymetrix HT Mouse Genome 430B Array 1434726_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 398632 - -
          ABI Mouse Genome Survey Microarray 738191 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3520324 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003520324 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1234738 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1234738 - -
          mouse-6 v1.1 (Illumina) scl50591.13.1_153-S - -
          Illumina Sentrix mouse-6.v1 bead array 3520324 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50591.13.1_153-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3520324 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1234738 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1234738 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline