Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00617420_s1
          See other CCDC184 GE Assays ›
          Gene Symbol:
          Ccdc184
          Gene Name:
          coiled-coil domain containing 184
          Gene Aliases:
          5630400A08, AI836003
          Chromosome Location:
          Chr.15: 98167806 - 98170134 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00617420_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccdc184
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177716.3 NP_808384.2 1 1102 81
          GenBank mRNA AK030707.1 - 1 647 81
          AK080988.1 - 1 1248 81
          BC090621.1 - 1 1012 6828730 81
          BC095968.1 - 1 582 6492111 81

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccdc184

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30473514 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30473515 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30473509 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30473516 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30473511 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30473513 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30473510 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30473517 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169227 AK080988.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 769558 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117038 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169227 AK080988.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473480 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473482 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30473499 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P117038 - -
          Whole Mouse Genome G4122A A_51_P147024 BC090621.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P147024 BC095968.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P147024 NM_177716.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P147024 AK030707.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107674 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P117038 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12437519 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12033046 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12437497 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12437505 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30473511 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30473515 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30473510 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30473516 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30473509 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30473514 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30473513 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30473517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473475 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11853694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11857062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473464 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12437523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11857063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473465 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12097955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11838216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12055988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11992914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12150704 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12312179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473467 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473481 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12055997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11838212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473495 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473503 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473492 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12033046 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12437497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473487 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473473 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12033064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473480 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473499 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10426583 - -
          Affymetrix HT MG-430 PM Array Plate 1435512_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1436099_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10426581 - -
          Mouse Genome 430 2.0 Array 1436099_PM_at - -
          Mouse Genome 430 2.0 Array 1435512_PM_at - -
          Mouse Genome 430 2.0 Array 1436099_at - -
          Mouse Genome 430 2.0 Array 1435512_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 239650_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 239650_at - -
          Affymetrix Mouse Clariom S Array PSR1500007633.mm.1 NM_177716.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500007633.mm.1 NM_177716.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500007633.mm.1 TC1500000971.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500007634.mm.1 NM_177716.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500007634.mm.1 NM_177716.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500007634.mm.1 TC1500000971.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500007632.mm.1 NM_177716.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500007632.mm.1 NM_177716.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500007632.mm.1 TC1500000971.mm.1 MTA TC
          Mouse Expression Set 430 1436099_at - -
          Mouse Expression Set 430 1435512_at - -
          Mouse Expression Set 430 1435512_PM_at - -
          Mouse Expression Set 430 1436099_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436099_at - -
          Affymetrix HT Mouse Genome 430B Array 1435512_at - -
          Affymetrix HT Mouse Genome 430B Array 1436099_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435512_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 309034 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3780735 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003780735 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2732079 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2732079 - -
          mouse-6 v1.1 (Illumina) scl47556.1.3_35-S - -
          Illumina Sentrix mouse-6.v1 bead array 3780735 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl47556.1.3_35-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3780735 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2732079 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2732079 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl47556.1.3_35-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline