Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00620585_m1
          See other ZFP641 GE Assays ›
          Gene Symbol:
          Zfp641
          Gene Name:
          zinc finger protein 641
          Gene Aliases:
          9930016F01Rik, Znf641
          Chromosome Location:
          Chr.15: 98285590 - 98296508 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00620585_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp641
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173769.3 NP_776130.1 3 - 4 328 72
          GenBank mRNA AK036826.1 - 3 - 4 329 72
          AK087530.1 - 3 - 4 376 72
          AK136402.1 - 3 - 4 406 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500002170.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp641

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          nucleic acid binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30474024 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30474023 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30474026 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30474025 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30474028 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30474027 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P392644 AK036826.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P392644 AK087530.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P392644 NM_173769.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P392644 AK036826.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P392644 AK087530.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P392644 NM_173769.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 8716 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 396076 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 4454 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 364668 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 407215 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 86737 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 67442 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146489 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140745 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P392644 AK036826.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P392644 AK087530.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P392644 NM_173769.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P392644 AK036826.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P392644 AK087530.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P392644 NM_173769.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474007 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08475965 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140745 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P146489 - -
          Whole Mouse Genome G4122A A_52_P392644 AK036826.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P392644 AK087530.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P392644 NM_173769.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P392644 AK036826.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P392644 AK087530.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P392644 NM_173769.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134231 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112328 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140745 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P146489 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12013002 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12444105 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12444092 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12444104 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12444095 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30474028 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30474025 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30474023 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30474026 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30474024 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30474027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12275127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11938920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474011 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12309456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12309463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12309469 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474046 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12463132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11849820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12450031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473958 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10432144 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432143 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432142 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432148 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432147 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432146 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432141 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432145 - -
          Affymetrix Mouse Gene 1.1 ST Array 10432139 - -
          Affymetrix Mouse Clariom S Array PSR1500017621.mm.1 NM_173769.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500017621.mm.1 NM_173769.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500017621.mm.1 TC1500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500017620.mm.1 NM_173769.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500017620.mm.1 NM_173769.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500017620.mm.1 TC1500002170.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 381063 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4060338 - -
          Illumina mouse-6 v1.0 expression beadchip 103990497 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004060338 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1256695 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256695 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2493040 - -
          mouse-6 v1.1 (Illumina) scl00239652.2_53-S - -
          mouse-6 v1.1 (Illumina) scl46772.7_573-S - -
          Illumina Sentrix mouse-6.v1 bead array 4060338 - -
          Illumina Sentrix mouse-6.v1 bead array 103990497 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00239652.2_53-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103990497 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4060338 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1256695 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1256695 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl00239652.2_53-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline