Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00622883_s1
          See other B330016D10RIK GE Assays ›
          Gene Symbol:
          B330016D10Rik
          Gene Name:
          RIKEN cDNA B330016D10 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 141546162 - 141548313 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00622883_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: B330016D10Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_030695.1 1 1371 100
          GenBank mRNA AK046111.1 - 1 1371 100
          AK046532.1 - 1 1367 100
          AK080800.1 - 1 1378 100
          AK080864.1 - 1 1347 100
          AK082239.1 - 1 749 100
          AK148126.1 - 1 719 100

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P305053 AK148126.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P305053 AK080800.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P305053 AK080864.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P305053 AK082239.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P305053 NR_030695.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P305053 AK148126.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P305053 AK080800.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P305053 AK080864.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P305053 AK082239.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P305053 NR_030695.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 496052 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 55755 - -
          Whole Mouse Genome G4122A A_52_P305053 AK148126.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P305053 AK080800.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P305053 AK080864.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P305053 AK082239.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P305053 NR_030695.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P305053 AK148126.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P305053 AK080800.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P305053 AK080864.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P305053 AK082239.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P305053 NR_030695.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P23364747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23364716 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03315153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23364721 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03315157 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03839812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23364730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23364725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23364727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23364717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23364745 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10510020 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510021 - -
          Affymetrix HT MG-430 PM Array Plate 1455492_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10510019 - -
          Mouse Genome 430 2.0 Array 1455492_at - -
          Mouse Genome 430 2.0 Array 1455492_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 320456_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 320456_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400014903.mm.1 NR_030695.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014903.mm.1 NR_030695.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014903.mm.1 TC0400001794.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400014897.mm.1 NR_030695.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014897.mm.1 NR_030695.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014897.mm.1 TC0400001794.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400014901.mm.1 NR_030695.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014901.mm.1 NR_030695.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014901.mm.1 TC0400001794.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400014899.mm.1 NR_030695.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014899.mm.1 NR_030695.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014899.mm.1 TC0400001794.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400014904.mm.1 NR_030695.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014904.mm.1 NR_030695.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014904.mm.1 TC0400001794.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400014902.mm.1 NR_030695.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014902.mm.1 NR_030695.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014902.mm.1 TC0400001794.mm.1 MTA TC
          Mouse Expression Set 430 1455492_at - -
          Mouse Expression Set 430 1455492_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455492_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455492_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 315811 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0