Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00625907_m1
          See other 4933411G11RIK GE Assays ›
          Gene Symbol:
          4933411G11Rik
          Gene Name:
          RIKEN cDNA 4933411G11Rik gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 143180924 - 143205069 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00625907_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4933411G11Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177880.4 NP_808548.1 2 - 3 610 79
          GenBank mRNA AK030175.1 - 2 - 3 611 79
          BC115568.1 - 2 - 3 143 40102090 79
          BC115569.1 - 2 - 3 232 40102091 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001725.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 4933411G11Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          regulation of protein kinase activity
          molecular_function
          protein kinase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P170565 AK030175.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P170565 NM_177880.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P170565 AK030175.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P170565 NM_177880.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 253863 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 63868 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 635613 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 25875 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 682954 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 186338 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599796 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 56094 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 587830 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 9322 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 482992 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 6767 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 15509 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 128304 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158483 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155743 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P170565 AK030175.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P170565 NM_177880.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P170565 AK030175.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P170565 NM_177880.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24155344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24155343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24155351 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24155350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24155353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24155358 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24155356 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24155354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24155337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24155347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24155348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24155345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24155359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24155361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24155336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24155335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24155357 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05774491 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155743 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05774500 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158483 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05774468 - -
          Whole Mouse Genome G4122A A_51_P170565 AK030175.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P170565 NM_177880.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P170565 AK030175.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P170565 NM_177880.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127639 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158483 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155743 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04763919 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04763922 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04763934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24155383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24155356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04601329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24155334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04898490 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04709817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24155350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04582743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04469021 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24155347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04709810 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04601331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24155335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24155336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24155353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04601332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24155349 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10527167 - -
          Affymetrix Mouse Gene 1.0 ST Array 10527165 - -
          Affymetrix Mouse Gene 1.0 ST Array 10527168 - -
          Affymetrix Mouse Gene 1.0 ST Array 10527170 - -
          Affymetrix Mouse Gene 1.0 ST Array 10527169 - -
          Affymetrix Mouse Gene 1.0 ST Array 10527171 - -
          Affymetrix Mouse Gene 1.0 ST Array 10527166 - -
          Affymetrix Mouse Gene 1.1 ST Array 10527164 - -
          Affymetrix Mouse Clariom S Array PSR0500015824.mm.1 TC0500001725.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015824.mm.1 NM_177880.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015824.mm.1 NM_177880.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015829.mm.1 TC0500001725.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015829.mm.1 NM_177880.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015829.mm.1 NM_177880.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015825.mm.1 TC0500001725.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015825.mm.1 NM_177880.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015825.mm.1 NM_177880.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015827.mm.1 TC0500001725.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015827.mm.1 NM_177880.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015827.mm.1 NM_177880.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015830.mm.1 TC0500001725.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015830.mm.1 NM_177880.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015830.mm.1 NM_177880.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015826.mm.1 TC0500001725.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015828.mm.1 TC0500001725.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015828.mm.1 NM_177880.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015828.mm.1 NM_177880.4 Refseq
          Murine Genome U74v2 Set 136099_PM_at - -
          Murine Genome U74v2 Set 167545_PM_r_at - -
          Murine Genome U74v2 Set 167545_r_at - -
          Murine Genome U74v2 Set 136099_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 744273 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6660452 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006660452 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2675932 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2675932 - -
          mouse-6 v1.1 (Illumina) scl27022.8.1_245-S - -
          Illumina Sentrix mouse-6.v1 bead array 6660452 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl27022.8.1_245-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6660452 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2675932 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2675932 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0