Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00626152_m1
          See other LCN10 GE Assays ›
          Gene Symbol:
          Lcn10
          Gene Name:
          lipocalin 10
          Gene Aliases:
          9230112J07Rik
          Chromosome Location:
          Chr.2: 25682726 - 25686081 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00626152_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lcn10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178036.4 NP_828875.1 1 - 2 161 61
          GenBank mRNA AF435737.1 - 1 - 2 161 61
          BC119103.1 - 1 - 2 129 8733852 61
          BC120657.1 - 1 - 2 129 40129580 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000430.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lcn10

          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343661 NM_178036.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343661 AF435737.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343661 AK020338.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343661 BC119103.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343661 BC120657.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343661 NM_178036.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343661 AF435737.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343661 AK020338.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343661 BC119103.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343661 BC120657.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 645966 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 824467 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131324 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343661 NM_178036.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343661 AF435737.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343661 AK020338.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343661 BC119103.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343661 BC120657.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343661 NM_178036.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343661 AF435737.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343661 AK020338.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343661 BC119103.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343661 BC120657.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073724 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073726 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073712 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073717 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073731 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131324 - -
          Whole Mouse Genome G4122A A_52_P343661 NM_178036.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343661 AF435737.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343661 AK020338.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343661 BC119103.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343661 BC120657.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343661 NM_178036.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343661 AF435737.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343661 AK020338.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343661 BC119103.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343661 BC120657.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128838 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131324 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01796770 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01796830 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01796780 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01796789 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01796828 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01796766 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01796788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073778 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01851259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01796797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01837360 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01796793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01796830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01227975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073780 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01796772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01796766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073751 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073783 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10470168 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470167 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470173 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470171 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470169 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470172 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470174 - -
          Affymetrix Mouse Gene 1.1 ST Array 10470166 - -
          Affymetrix Mouse Clariom S Array PSR0200002645.mm.1 NM_178036.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002645.mm.1 TC0200000430.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200002644.mm.1 NM_178036.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002644.mm.1 TC0200000430.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200002643.mm.1 NM_178036.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002643.mm.1 TC0200000430.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 656460 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1740048 - -
          Illumina mouse-6 v1.0 expression beadchip 1740048 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001740048 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1213917 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1213917 - -
          mouse-6 v1.1 (Illumina) scl21167.6.1_82-S - -
          Illumina Sentrix mouse-6.v1 bead array 1740048 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21167.6.1_82-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1740048 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1213917 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1213917 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline