Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00652703_s1
          See other VMN1R226 GE Assays ›
          Gene Symbol:
          Vmn1r226
          Gene Name:
          vomeronasal 1 receptor 226
          Gene Aliases:
          V1re2
          Chromosome Location:
          Chr.17: 20687508 - 20688404 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00652703_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r226
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134191.1 NP_598952.1 1 676 150
          GenBank mRNA BC119429.1 - 1 674 150
          BC119430.1 - 1 676 40040474 150

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r226

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          response to pheromone
          pheromone binding
          pheromone receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P484704 BC119429.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P484704 BC119430.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P484704 NM_134191.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P484704 BC119429.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P484704 BC119430.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P484704 NM_134191.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P169605 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P484704 BC119429.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P484704 BC119430.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P484704 NM_134191.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P484704 BC119429.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P484704 BC119430.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P484704 NM_134191.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083578 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083604 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P169605 - -
          Whole Mouse Genome G4122A A_52_P484704 BC119429.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P484704 BC119430.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P484704 NM_134191.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P484704 BC119429.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P484704 BC119430.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P484704 NM_134191.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139990 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P169605 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13384255 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13411775 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13411758 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13411759 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13411760 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13411756 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13411768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13570231 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13384257 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083629 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083636 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442146 - -
          Affymetrix Mouse Gene 1.1 ST Array 10442145 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 774302 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1400593 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001400593 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1221165 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1221165 - -
          mouse-6 v1.1 (Illumina) scl8636.1.1_326-S - -
          Illumina Sentrix mouse-6.v1 bead array 1400593 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl8636.1.1_326-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1400593 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2823366 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2823366 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline